Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 143215344-143215585 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPIHBP1
Entrez Gene ID
338328
Gene Name
glycosylphosphatidylinositol anchored high density lipoprotein binding protein 1
Gene Aliases
GPI-HBP1, HYPL1D
UniGene
Hs.426410
Gene Chromosome Location
Chr.8: 143213193-143217170 on Build GRCh38
Transcripts
NM_001301772.1 NM_178172.5

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5408951 c.483C>T p.S161S Chr.8: 143215446-143215446 on Build GRCh38
5011960 c.537G>A p.M179I Chr.8: 143215500-143215500 on Build GRCh38
6180243 c.434G>T p.R145L Chr.8: 143215397-143215397 on Build GRCh38
6180242 c.469C>G p.P157A Chr.8: 143215432-143215432 on Build GRCh38
3645997 c.482C>T p.S161F Chr.8: 143215445-143215445 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145844329 Chr.8: 143215486-143215486 on Build GRCh38
rs770221336 Chr.8: 143215397-143215397 on Build GRCh38
rs773445832 Chr.8: 143215504-143215504 on Build GRCh38
rs774794706 Chr.8: 143215547-143215547 on Build GRCh38
rs752724041 Chr.8: 143215442-143215442 on Build GRCh38
rs772237000 Chr.8: 143215553-143215553 on Build GRCh38
rs966148826 Chr.8: 143215427-143215427 on Build GRCh38
rs749587698 Chr.8: 143215535-143215535 on Build GRCh38
rs746715981 Chr.8: 143215381-143215381 on Build GRCh38
rs78367243 Chr.8: 143215394-143215394 on Build GRCh38
rs756748449 Chr.8: 143215524-143215524 on Build GRCh38
rs767655645 Chr.8: 143215501-143215501 on Build GRCh38
rs780533342 Chr.8: 143215527-143215527 on Build GRCh38
rs762093328 Chr.8: 143215435-143215435 on Build GRCh38
rs780464769 Chr.8: 143215533-143215533 on Build GRCh38
rs374384937 Chr.8: 143215432-143215432 on Build GRCh38
rs756979938 Chr.8: 143215458-143215458 on Build GRCh38
rs761823083 Chr.8: 143215496-143215496 on Build GRCh38
rs202049123 Chr.8: 143215507-143215507 on Build GRCh38
rs1010225016 Chr.8: 143215380-143215380 on Build GRCh38
rs371930918 Chr.8: 143215517-143215517 on Build GRCh38
rs573899249 Chr.8: 143215531-143215531 on Build GRCh38
rs376669795 Chr.8: 143215440-143215440 on Build GRCh38
rs866602255 Chr.8: 143215379-143215379 on Build GRCh38
rs552438640 Chr.8: 143215556-143215556 on Build GRCh38
rs770702398 Chr.8: 143215392-143215392 on Build GRCh38
rs200984081 Chr.8: 143215523-143215523 on Build GRCh38
rs373297994 Chr.8: 143215447-143215447 on Build GRCh38
rs746247359 Chr.8: 143215459-143215459 on Build GRCh38
rs776022117 Chr.8: 143215401-143215401 on Build GRCh38
rs745582068 Chr.8: 143215528-143215528 on Build GRCh38
rs747105275 Chr.8: 143215493-143215493 on Build GRCh38
rs767103438 Chr.8: 143215430-143215431 on Build GRCh38
rs767716301 Chr.8: 143215436-143215436 on Build GRCh38
rs564622687 Chr.8: 143215438-143215438 on Build GRCh38
rs756773250 Chr.8: 143215424-143215425 on Build GRCh38
rs373232320 Chr.8: 143215439-143215439 on Build GRCh38
rs775697635 Chr.8: 143215467-143215467 on Build GRCh38
rs746452521 Chr.8: 143215456-143215456 on Build GRCh38
rs548469693 Chr.8: 143215489-143215489 on Build GRCh38
rs751092284 Chr.8: 143215423-143215424 on Build GRCh38
rs763417538 Chr.8: 143215415-143215415 on Build GRCh38
rs563623646 Chr.8: 143215514-143215514 on Build GRCh38
rs759883512 Chr.8: 143215396-143215396 on Build GRCh38
rs370155321 Chr.8: 143215446-143215446 on Build GRCh38
rs749855006 Chr.8: 143215485-143215485 on Build GRCh38
rs559317477 Chr.8: 143215461-143215461 on Build GRCh38
rs587777642 Chr.8: 143215380-143215396 on Build GRCh38
rs139851434 Chr.8: 143215428-143215428 on Build GRCh38
rs1022802744 Chr.8: 143215555-143215555 on Build GRCh38
rs149828979 Chr.8: 143215429-143215429 on Build GRCh38
rs769165369 Chr.8: 143215541-143215541 on Build GRCh38
rs748262356 Chr.8: 143215551-143215551 on Build GRCh38
rs750040173 Chr.8: 143215519-143215519 on Build GRCh38

View Full Product Details