Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 140128436-140128665 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IGIP
Entrez Gene ID
492311
Gene Name
IgA inducing protein
Gene Aliases
C5orf53
UniGene
Hs.696360
Gene Chromosome Location
Chr.5: 140125936-140128806 on Build GRCh38
Transcripts
NM_001007189.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3609292 c.53C>T p.T18I Chr.5: 140128529-140128529 on Build GRCh38
3609293 c.97C>T p.P33S Chr.5: 140128573-140128573 on Build GRCh38
3238361 c.51T>A p.I17I Chr.5: 140128527-140128527 on Build GRCh38
3238363 c.116A>C p.N39T Chr.5: 140128592-140128592 on Build GRCh38
5162963 c.133C>T p.R45W Chr.5: 140128609-140128609 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150564736 Chr.5: 140128504-140128504 on Build GRCh38
rs141663065 Chr.5: 140128479-140128479 on Build GRCh38
rs778332752 Chr.5: 140128496-140128496 on Build GRCh38
rs771084947 Chr.5: 140128550-140128550 on Build GRCh38
rs575850066 Chr.5: 140128540-140128540 on Build GRCh38
rs773133022 Chr.5: 140128533-140128533 on Build GRCh38
rs780772095 Chr.5: 140128517-140128517 on Build GRCh38
rs754538078 Chr.5: 140128610-140128610 on Build GRCh38
rs115235886 Chr.5: 140128528-140128528 on Build GRCh38
rs542562858 Chr.5: 140128495-140128495 on Build GRCh38
rs939807945 Chr.5: 140128552-140128552 on Build GRCh38
rs765769869 Chr.5: 140128602-140128602 on Build GRCh38
rs139544518 Chr.5: 140128523-140128523 on Build GRCh38
rs199981499 Chr.5: 140128494-140128494 on Build GRCh38
rs199527880 Chr.5: 140128567-140128567 on Build GRCh38
rs769487440 Chr.5: 140128531-140128531 on Build GRCh38
rs530033348 Chr.5: 140128515-140128515 on Build GRCh38
rs746567746 Chr.5: 140128514-140128514 on Build GRCh38
rs369243227 Chr.5: 140128490-140128490 on Build GRCh38
rs757735362 Chr.5: 140128592-140128592 on Build GRCh38
rs773079358 Chr.5: 140128477-140128477 on Build GRCh38
rs372747459 Chr.5: 140128485-140128487 on Build GRCh38
rs759859798 Chr.5: 140128487-140128487 on Build GRCh38
rs775635990 Chr.5: 140128572-140128572 on Build GRCh38
rs373649776 Chr.5: 140128573-140128573 on Build GRCh38
rs919803623 Chr.5: 140128593-140128593 on Build GRCh38
rs779574619 Chr.5: 140128507-140128507 on Build GRCh38
rs188052223 Chr.5: 140128626-140128626 on Build GRCh38
rs376262537 Chr.5: 140128576-140128576 on Build GRCh38
rs201733032 Chr.5: 140128609-140128609 on Build GRCh38
rs75647802 Chr.5: 140128569-140128569 on Build GRCh38
rs780838079 Chr.5: 140128643-140128643 on Build GRCh38
rs757841420 Chr.5: 140128505-140128505 on Build GRCh38
rs747759546 Chr.5: 140128645-140128645 on Build GRCh38
rs764373966 Chr.5: 140128575-140128575 on Build GRCh38
rs763031143 Chr.5: 140128478-140128478 on Build GRCh38

View Full Product Details