Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 7727616-7727889 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITIH2
Entrez Gene ID
3698
Gene Name
inter-alpha-trypsin inhibitor heavy chain 2
Gene Aliases
H2P, SHAP
UniGene
Hs.75285
Gene Chromosome Location
Chr.10: 7703273-7749520 on Build GRCh38
Transcripts
NM_002216.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4461057 c.1191C>T p.F397F Chr.10: 7727740-7727740 on Build GRCh38
3440718 c.1234T>A p.S412T Chr.10: 7727783-7727783 on Build GRCh38
4015973 c.1279G>T p.G427C Chr.10: 7727828-7727828 on Build GRCh38
920583 c.1277_1278insGGGC p.E428fs*13 Chr.10: 7727826-7727827 on Build GRCh38
3440717 c.1216G>T p.G406* Chr.10: 7727765-7727765 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs767782376 Chr.10: 7727705-7727705 on Build GRCh38
rs760534201 Chr.10: 7727671-7727671 on Build GRCh38
rs778103865 Chr.10: 7727833-7727833 on Build GRCh38
rs201966331 Chr.10: 7727819-7727819 on Build GRCh38
rs548587256 Chr.10: 7727717-7727717 on Build GRCh38
rs770825404 Chr.10: 7727755-7727755 on Build GRCh38
rs137948551 Chr.10: 7727716-7727716 on Build GRCh38
rs766492548 Chr.10: 7727678-7727678 on Build GRCh38
rs146198523 Chr.10: 7727703-7727703 on Build GRCh38
rs759507283 Chr.10: 7727772-7727772 on Build GRCh38
rs41290289 Chr.10: 7727843-7727843 on Build GRCh38
rs374140482 Chr.10: 7727854-7727854 on Build GRCh38
rs939387529 Chr.10: 7727835-7727835 on Build GRCh38
rs200682259 Chr.10: 7727781-7727781 on Build GRCh38
rs2277226 Chr.10: 7727641-7727641 on Build GRCh38
rs923938714 Chr.10: 7727816-7727816 on Build GRCh38
rs776731372 Chr.10: 7727768-7727768 on Build GRCh38
rs759165056 Chr.10: 7727680-7727680 on Build GRCh38
rs370071328 Chr.10: 7727853-7727853 on Build GRCh38
rs139479101 Chr.10: 7727786-7727786 on Build GRCh38
rs141611690 Chr.10: 7727735-7727735 on Build GRCh38
rs752323725 Chr.10: 7727699-7727699 on Build GRCh38
rs373326533 Chr.10: 7727662-7727662 on Build GRCh38
rs780591182 Chr.10: 7727722-7727722 on Build GRCh38
rs368912521 Chr.10: 7727814-7727814 on Build GRCh38
rs141792551 Chr.10: 7727668-7727668 on Build GRCh38
rs530159598 Chr.10: 7727691-7727691 on Build GRCh38
rs377542536 Chr.10: 7727679-7727679 on Build GRCh38
rs201613595 Chr.10: 7727656-7727656 on Build GRCh38
rs768005405 Chr.10: 7727861-7727861 on Build GRCh38
rs758379393 Chr.10: 7727827-7727827 on Build GRCh38
rs762562641 Chr.10: 7727860-7727860 on Build GRCh38
rs764653891 Chr.10: 7727776-7727776 on Build GRCh38
rs150505580 Chr.10: 7727753-7727753 on Build GRCh38
rs200461639 Chr.10: 7727655-7727655 on Build GRCh38
rs781750816 Chr.10: 7727667-7727667 on Build GRCh38
rs374174475 Chr.10: 7727669-7727669 on Build GRCh38
rs747178163 Chr.10: 7727839-7727839 on Build GRCh38
rs775003689 Chr.10: 7727782-7727782 on Build GRCh38
rs762942003 Chr.10: 7727727-7727728 on Build GRCh38
rs370458835 Chr.10: 7727729-7727729 on Build GRCh38
rs200443041 Chr.10: 7727730-7727730 on Build GRCh38
rs756497031 Chr.10: 7727791-7727791 on Build GRCh38
rs534842022 Chr.10: 7727822-7727822 on Build GRCh38
rs769267995 Chr.10: 7727776-7727776 on Build GRCh38
rs267602583 Chr.10: 7727810-7727810 on Build GRCh38
rs775282561 Chr.10: 7727859-7727859 on Build GRCh38
rs748332598 Chr.10: 7727738-7727738 on Build GRCh38
rs751151848 Chr.10: 7727790-7727790 on Build GRCh38
rs755420130 Chr.10: 7727812-7727812 on Build GRCh38
rs377531123 Chr.10: 7727663-7727663 on Build GRCh38
rs377177337 Chr.10: 7727846-7727846 on Build GRCh38
rs766693296 Chr.10: 7727799-7727799 on Build GRCh38
rs754351617 Chr.10: 7727803-7727803 on Build GRCh38
rs138692753 Chr.10: 7727785-7727785 on Build GRCh38
rs772398280 Chr.10: 7727739-7727739 on Build GRCh38
rs773304469 Chr.10: 7727749-7727749 on Build GRCh38

View Full Product Details