Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 19227280-19227505 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMC1
Entrez Gene ID
23065
Gene Name
ER membrane protein complex subunit 1
Gene Aliases
CAVIPMR, KIAA0090
UniGene
Hs.439200
Gene Chromosome Location
Chr.1: 19215664-19251559 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 XM_005245787.1 XM_005245788.1 NR_135114.1 NR_135115.1

Gene Symbol
LOC101927895
Entrez Gene ID
101927895
Gene Name
uncharacterized LOC101927895
Gene Aliases
-
UniGene
Hs.660534
Gene Chromosome Location
Chr.1: 19210386-19240704 on Build GRCh38
Transcripts
NM_001271427.1 NM_001271428.1 NM_001271429.1 NM_015047.2 XM_005245787.1 XM_005245788.1 NR_135114.1 NR_135115.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6226258 c.2141G>A p.R714H Chr.1: 19227374-19227374 on Build GRCh38
4026427 c.2122G>A p.V708I Chr.1: 19227393-19227393 on Build GRCh38
1211831 c.2171G>A p.R724H Chr.1: 19227344-19227344 on Build GRCh38
6653601 c.2185C>T p.R729C Chr.1: 19227330-19227330 on Build GRCh38
240339 c.2140C>T p.R714C Chr.1: 19227375-19227375 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751157950 Chr.1: 19227322-19227322 on Build GRCh38
rs766121505 Chr.1: 19227319-19227319 on Build GRCh38
rs745907513 Chr.1: 19227463-19227463 on Build GRCh38
rs749944857 Chr.1: 19227314-19227314 on Build GRCh38
rs138291577 Chr.1: 19227476-19227476 on Build GRCh38
rs775852029 Chr.1: 19227376-19227376 on Build GRCh38
rs765783782 Chr.1: 19227397-19227397 on Build GRCh38
rs756136017 Chr.1: 19227344-19227344 on Build GRCh38
rs139714938 Chr.1: 19227393-19227393 on Build GRCh38
rs753806521 Chr.1: 19227459-19227459 on Build GRCh38
rs199561891 Chr.1: 19227398-19227398 on Build GRCh38
rs188945281 Chr.1: 19227399-19227399 on Build GRCh38
rs373742422 Chr.1: 19227475-19227475 on Build GRCh38
rs757833511 Chr.1: 19227316-19227316 on Build GRCh38
rs767221166 Chr.1: 19227402-19227402 on Build GRCh38
rs777919151 Chr.1: 19227454-19227454 on Build GRCh38
rs755969608 Chr.1: 19227445-19227445 on Build GRCh38
rs747905122 Chr.1: 19227334-19227334 on Build GRCh38
rs749449853 Chr.1: 19227361-19227361 on Build GRCh38
rs778894345 Chr.1: 19227462-19227462 on Build GRCh38
rs770906017 Chr.1: 19227366-19227366 on Build GRCh38
rs896315803 Chr.1: 19227374-19227374 on Build GRCh38
rs1052665530 Chr.1: 19227360-19227360 on Build GRCh38
rs780181389 Chr.1: 19227479-19227479 on Build GRCh38
rs143090840 Chr.1: 19227321-19227321 on Build GRCh38
rs774684957 Chr.1: 19227368-19227368 on Build GRCh38
rs962063326 Chr.1: 19227464-19227464 on Build GRCh38
rs781072054 Chr.1: 19227410-19227410 on Build GRCh38
rs760968223 Chr.1: 19227387-19227387 on Build GRCh38
rs370661581 Chr.1: 19227394-19227394 on Build GRCh38
rs754850089 Chr.1: 19227327-19227327 on Build GRCh38
rs746049382 Chr.1: 19227371-19227371 on Build GRCh38
rs777786640 Chr.1: 19227345-19227345 on Build GRCh38
rs1005818316 Chr.1: 19227426-19227426 on Build GRCh38
rs772294287 Chr.1: 19227375-19227375 on Build GRCh38
rs376593702 Chr.1: 19227330-19227330 on Build GRCh38

View Full Product Details