Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 38982856-38983129 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HTRA4
Entrez Gene ID
203100
Gene Name
HtrA serine peptidase 4
Gene Aliases
-
UniGene
Hs.661014
Gene Chromosome Location
Chr.8: 38974149-38988662 on Build GRCh38
Transcripts
NM_153692.3 XM_011544431.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5463726 c.1173-5C>A p.? Chr.8: 38982948-38982948 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs776587945 Chr.8: 38983031-38983031 on Build GRCh38
rs201875131 Chr.8: 38983073-38983073 on Build GRCh38
rs375362815 Chr.8: 38983071-38983071 on Build GRCh38
rs760658071 Chr.8: 38983008-38983008 on Build GRCh38
rs969995861 Chr.8: 38983102-38983102 on Build GRCh38
rs201171544 Chr.8: 38982956-38982956 on Build GRCh38
rs769106946 Chr.8: 38982916-38982916 on Build GRCh38
rs776546047 Chr.8: 38983020-38983020 on Build GRCh38
rs929545309 Chr.8: 38982987-38982987 on Build GRCh38
rs17854928 Chr.8: 38983026-38983026 on Build GRCh38
rs187664938 Chr.8: 38983000-38983000 on Build GRCh38
rs372581115 Chr.8: 38982929-38982929 on Build GRCh38
rs764168574 Chr.8: 38983015-38983015 on Build GRCh38
rs747534255 Chr.8: 38982909-38982909 on Build GRCh38
rs754738686 Chr.8: 38982960-38982961 on Build GRCh38
rs34058763 Chr.8: 38983013-38983014 on Build GRCh38
rs560605138 Chr.8: 38983040-38983040 on Build GRCh38
rs758287812 Chr.8: 38983027-38983027 on Build GRCh38
rs774460402 Chr.8: 38983081-38983081 on Build GRCh38
rs376879603 Chr.8: 38982999-38982999 on Build GRCh38
rs750029991 Chr.8: 38983035-38983035 on Build GRCh38
rs375670613 Chr.8: 38982904-38982904 on Build GRCh38
rs753736580 Chr.8: 38982956-38982957 on Build GRCh38
rs200212415 Chr.8: 38983069-38983069 on Build GRCh38
rs771126408 Chr.8: 38983074-38983074 on Build GRCh38
rs755484999 Chr.8: 38983054-38983054 on Build GRCh38
rs1042790607 Chr.8: 38982920-38982920 on Build GRCh38
rs7814264 Chr.8: 38983072-38983072 on Build GRCh38
rs765903899 Chr.8: 38983049-38983049 on Build GRCh38
rs370207714 Chr.8: 38983083-38983083 on Build GRCh38
rs763089883 Chr.8: 38982985-38982985 on Build GRCh38
rs545710209 Chr.8: 38983006-38983006 on Build GRCh38
rs369022593 Chr.8: 38982923-38982923 on Build GRCh38
rs770165390 Chr.8: 38982948-38982948 on Build GRCh38

View Full Product Details