Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 139167108-139167351 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECA
Entrez Gene ID
51696
Gene Name
hdc homolog, cell cycle regulator
Gene Aliases
HDC, HDCL, HHDC, dJ225E12.1
UniGene
Hs.197644
Gene Chromosome Location
Chr.6: 139135112-139180809 on Build GRCh38
Transcripts
NM_016217.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3016439 c.1193G>C p.C398S Chr.6: 139167205-139167205 on Build GRCh38
4155987 c.1276G>T p.D426Y Chr.6: 139167288-139167288 on Build GRCh38
6678665 c.1285G>A p.E429K Chr.6: 139167297-139167297 on Build GRCh38
3016435 c.1105G>C p.E369Q Chr.6: 139167117-139167117 on Build GRCh38
5908627 c.1265C>T p.P422L Chr.6: 139167277-139167277 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs367751153 Chr.6: 139167224-139167224 on Build GRCh38
rs1042616058 Chr.6: 139167277-139167277 on Build GRCh38
rs373649622 Chr.6: 139167165-139167165 on Build GRCh38
rs950626351 Chr.6: 139167194-139167194 on Build GRCh38
rs747284214 Chr.6: 139167173-139167173 on Build GRCh38
rs138982846 Chr.6: 139167188-139167188 on Build GRCh38
rs142228127 Chr.6: 139167223-139167223 on Build GRCh38
rs532956161 Chr.6: 139167158-139167158 on Build GRCh38
rs774887565 Chr.6: 139167227-139167227 on Build GRCh38
rs983283988 Chr.6: 139167197-139167197 on Build GRCh38
rs749296857 Chr.6: 139167317-139167317 on Build GRCh38
rs945602715 Chr.6: 139167249-139167249 on Build GRCh38
rs145909139 Chr.6: 139167230-139167230 on Build GRCh38
rs917078350 Chr.6: 139167209-139167209 on Build GRCh38
rs781452221 Chr.6: 139167185-139167185 on Build GRCh38
rs145108878 Chr.6: 139167170-139167170 on Build GRCh38
rs374585437 Chr.6: 139167296-139167296 on Build GRCh38
rs749178753 Chr.6: 139167217-139167217 on Build GRCh38
rs766148085 Chr.6: 139167171-139167171 on Build GRCh38
rs201702729 Chr.6: 139167248-139167248 on Build GRCh38
rs756285622 Chr.6: 139167302-139167302 on Build GRCh38
rs375410347 Chr.6: 139167281-139167281 on Build GRCh38
rs34059634 Chr.6: 139167236-139167236 on Build GRCh38
rs774114464 Chr.6: 139167220-139167220 on Build GRCh38
rs562355741 Chr.6: 139167172-139167172 on Build GRCh38
rs751931133 Chr.6: 139167278-139167278 on Build GRCh38
rs750992864 Chr.6: 139167237-139167237 on Build GRCh38
rs541841684 Chr.6: 139167198-139167198 on Build GRCh38
rs551402071 Chr.6: 139167242-139167242 on Build GRCh38
rs777701910 Chr.6: 139167306-139167306 on Build GRCh38
rs773136893 Chr.6: 139167215-139167215 on Build GRCh38
rs370753473 Chr.6: 139167137-139167137 on Build GRCh38
rs138490244 Chr.6: 139167321-139167321 on Build GRCh38
rs770770683 Chr.6: 139167219-139167219 on Build GRCh38
rs868064784 Chr.6: 139167309-139167309 on Build GRCh38
rs35480523 Chr.6: 139167310-139167311 on Build GRCh38
rs760161738 Chr.6: 139167229-139167229 on Build GRCh38
rs927899185 Chr.6: 139167270-139167270 on Build GRCh38
rs200417003 Chr.6: 139167174-139167174 on Build GRCh38

View Full Product Details