Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45328089-45328328 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPDL
Entrez Gene ID
84842
Gene Name
4-hydroxyphenylpyruvate dioxygenase like
Gene Aliases
4-HPPD-L, GLOXD1
UniGene
Hs.162717
Gene Chromosome Location
Chr.1: 45326873-45328675 on Build GRCh38
Transcripts
NM_032756.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
7371452 c.964C>G p.L322V Chr.1: 45328112-45328112 on Build GRCh38
5904516 c.1094C>A p.S365Y Chr.1: 45328242-45328242 on Build GRCh38
3490220 c.996T>C p.T332T Chr.1: 45328144-45328144 on Build GRCh38
4418051 c.1065A>G p.R355R Chr.1: 45328213-45328213 on Build GRCh38
3934754 c.1022T>A p.I341N Chr.1: 45328170-45328170 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs369195593 Chr.1: 45328143-45328143 on Build GRCh38
rs748643827 Chr.1: 45328124-45328124 on Build GRCh38
rs368714969 Chr.1: 45328170-45328170 on Build GRCh38
rs1023864609 Chr.1: 45328249-45328249 on Build GRCh38
rs753509244 Chr.1: 45328207-45328207 on Build GRCh38
rs779698378 Chr.1: 45328225-45328225 on Build GRCh38
rs750059836 Chr.1: 45328214-45328214 on Build GRCh38
rs748733599 Chr.1: 45328228-45328228 on Build GRCh38
rs774472385 Chr.1: 45328260-45328260 on Build GRCh38
rs756946946 Chr.1: 45328212-45328212 on Build GRCh38
rs202009475 Chr.1: 45328300-45328300 on Build GRCh38
rs376654496 Chr.1: 45328254-45328254 on Build GRCh38
rs763244013 Chr.1: 45328141-45328141 on Build GRCh38
rs778216284 Chr.1: 45328229-45328229 on Build GRCh38
rs373876225 Chr.1: 45328176-45328176 on Build GRCh38
rs902898316 Chr.1: 45328199-45328199 on Build GRCh38
rs899771467 Chr.1: 45328157-45328157 on Build GRCh38
rs79702595 Chr.1: 45328213-45328213 on Build GRCh38
rs749570154 Chr.1: 45328235-45328235 on Build GRCh38
rs1042688354 Chr.1: 45328192-45328192 on Build GRCh38
rs775541131 Chr.1: 45328307-45328307 on Build GRCh38
rs145372387 Chr.1: 45328161-45328161 on Build GRCh38
rs148694275 Chr.1: 45328165-45328165 on Build GRCh38
rs368590349 Chr.1: 45328281-45328281 on Build GRCh38
rs764014580 Chr.1: 45328184-45328184 on Build GRCh38
rs781774728 Chr.1: 45328123-45328123 on Build GRCh38
rs770342580 Chr.1: 45328127-45328127 on Build GRCh38
rs748684818 Chr.1: 45328223-45328231 on Build GRCh38
rs758032205 Chr.1: 45328223-45328223 on Build GRCh38
rs868013173 Chr.1: 45328153-45328154 on Build GRCh38
rs760590250 Chr.1: 45328180-45328180 on Build GRCh38
rs773447476 Chr.1: 45328133-45328133 on Build GRCh38

View Full Product Details