Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 33205066-33205331 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HSD17B8
Entrez Gene ID
7923
Gene Name
hydroxysteroid 17-beta dehydrogenase 8
Gene Aliases
D6S2245E, FABG, FABGL, H2-KE6, HKE6, KE6, RING2, SDR30C1, dJ1033B10.9
UniGene
Hs.415058
Gene Chromosome Location
Chr.6: 33204637-33206831 on Build GRCh38
Transcripts
NM_014234.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4886974 c.235G>T p.A79S Chr.6: 33205084-33205084 on Build GRCh38
6682386 c.239G>T p.R80M Chr.6: 33205088-33205088 on Build GRCh38
421373 c.281C>G p.S94C Chr.6: 33205231-33205231 on Build GRCh38
4971102 c.280T>C p.S94P Chr.6: 33205230-33205230 on Build GRCh38
6203988 c.323A>T p.Q108L Chr.6: 33205273-33205273 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs370659717 Chr.6: 33205116-33205116 on Build GRCh38
rs191965195 Chr.6: 33205218-33205218 on Build GRCh38
rs759065768 Chr.6: 33205248-33205248 on Build GRCh38
rs764913157 Chr.6: 33205175-33205175 on Build GRCh38
rs779093965 Chr.6: 33205215-33205215 on Build GRCh38
rs574238516 Chr.6: 33205165-33205165 on Build GRCh38
rs749419029 Chr.6: 33205125-33205125 on Build GRCh38
rs773720520 Chr.6: 33205227-33205227 on Build GRCh38
rs764196969 Chr.6: 33205205-33205205 on Build GRCh38
rs750720359 Chr.6: 33205208-33205209 on Build GRCh38
rs759072636 Chr.6: 33205169-33205169 on Build GRCh38
rs110662 Chr.6: 33205155-33205155 on Build GRCh38
rs780331085 Chr.6: 33205118-33205118 on Build GRCh38
rs920242574 Chr.6: 33205224-33205224 on Build GRCh38
rs763927121 Chr.6: 33205271-33205271 on Build GRCh38
rs777835548 Chr.6: 33205102-33205102 on Build GRCh38
rs365339 Chr.6: 33205128-33205128 on Build GRCh38
rs144457204 Chr.6: 33205261-33205261 on Build GRCh38
rs556499620 Chr.6: 33205262-33205262 on Build GRCh38
rs138352449 Chr.6: 33205248-33205250 on Build GRCh38
rs751543629 Chr.6: 33205272-33205272 on Build GRCh38
rs779112327 Chr.6: 33205299-33205299 on Build GRCh38
rs755080187 Chr.6: 33205292-33205292 on Build GRCh38
rs769762232 Chr.6: 33205115-33205115 on Build GRCh38
rs757456441 Chr.6: 33205184-33205185 on Build GRCh38
rs755465645 Chr.6: 33205260-33205263 on Build GRCh38
rs758449941 Chr.6: 33205092-33205092 on Build GRCh38
rs201761806 Chr.6: 33205190-33205190 on Build GRCh38
rs141630682 Chr.6: 33205223-33205223 on Build GRCh38
rs756238441 Chr.6: 33205211-33205211 on Build GRCh38
rs776370953 Chr.6: 33205168-33205168 on Build GRCh38
rs751477353 Chr.6: 33205208-33205208 on Build GRCh38
rs749352910 Chr.6: 33205212-33205212 on Build GRCh38
rs753920057 Chr.6: 33205284-33205284 on Build GRCh38
rs749757690 Chr.6: 33205247-33205248 on Build GRCh38
rs376588362 Chr.6: 33205275-33205275 on Build GRCh38
rs758257314 Chr.6: 33205191-33205191 on Build GRCh38
rs770507785 Chr.6: 33205233-33205233 on Build GRCh38
rs751333450 Chr.6: 33205110-33205111 on Build GRCh38
rs776045109 Chr.6: 33205243-33205243 on Build GRCh38
rs767600140 Chr.6: 33205185-33205185 on Build GRCh38
rs746006075 Chr.6: 33205114-33205114 on Build GRCh38
rs756246487 Chr.6: 33205218-33205219 on Build GRCh38
rs774735143 Chr.6: 33205135-33205135 on Build GRCh38
rs772625395 Chr.6: 33205143-33205143 on Build GRCh38
rs767462927 Chr.6: 33205283-33205283 on Build GRCh38

View Full Product Details