Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 101915232-101915471 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANGPTL5
Entrez Gene ID
253935
Gene Name
angiopoietin like 5
Gene Aliases
-
UniGene
Hs.318370
Gene Chromosome Location
Chr.11: 101890674-101916522 on Build GRCh38
Transcripts
NM_178127.4 XM_011542735.2 XM_017017466.1 NM_020802.3 XM_005271627.4

Gene Symbol
CEP126
Entrez Gene ID
57562
Gene Name
centrosomal protein 126
Gene Aliases
KIAA1377
UniGene
Hs.156352
Gene Chromosome Location
Chr.11: 101915015-102001065 on Build GRCh38
Transcripts
NM_178127.4 XM_011542735.2 XM_017017466.1 NM_020802.3 XM_005271627.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3935276 c.38G>A p.G13E Chr.11: 101915322-101915322 on Build GRCh38
7176965 c.59C>T p.S20L Chr.11: 101915343-101915343 on Build GRCh38
6694002 c.65A>T p.N22I Chr.11: 101915349-101915349 on Build GRCh38
6067035 c.96G>C p.E32D Chr.11: 101915380-101915380 on Build GRCh38
1745999 c.69C>T p.L23L Chr.11: 101915353-101915353 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs146972831 Chr.11: 101915360-101915360 on Build GRCh38
rs536297070 Chr.11: 101915395-101915395 on Build GRCh38
rs779715132 Chr.11: 101915375-101915376 on Build GRCh38
rs988980652 Chr.11: 101915422-101915422 on Build GRCh38
rs200573063 Chr.11: 101915364-101915364 on Build GRCh38
rs150590684 Chr.11: 101915318-101915318 on Build GRCh38
rs149781214 Chr.11: 101915289-101915289 on Build GRCh38
rs951445862 Chr.11: 101915257-101915257 on Build GRCh38
rs776490810 Chr.11: 101915286-101915286 on Build GRCh38
rs1001644314 Chr.11: 101915346-101915346 on Build GRCh38
rs148913485 Chr.11: 101915309-101915309 on Build GRCh38
rs181761204 Chr.11: 101915317-101915317 on Build GRCh38
rs754372719 Chr.11: 101915444-101915444 on Build GRCh38
rs769861010 Chr.11: 101915296-101915296 on Build GRCh38
rs934091173 Chr.11: 101915251-101915251 on Build GRCh38
rs1039000697 Chr.11: 101915280-101915280 on Build GRCh38
rs374652030 Chr.11: 101915256-101915256 on Build GRCh38
rs759755371 Chr.11: 101915316-101915316 on Build GRCh38
rs749291141 Chr.11: 101915416-101915416 on Build GRCh38
rs935008107 Chr.11: 101915308-101915308 on Build GRCh38
rs758032724 Chr.11: 101915269-101915269 on Build GRCh38
rs749850977 Chr.11: 101915324-101915324 on Build GRCh38
rs757955825 Chr.11: 101915343-101915343 on Build GRCh38
rs746741985 Chr.11: 101915285-101915285 on Build GRCh38
rs762312229 Chr.11: 101915445-101915445 on Build GRCh38
rs201779479 Chr.11: 101915302-101915302 on Build GRCh38
rs865798047 Chr.11: 101915361-101915361 on Build GRCh38
rs774381609 Chr.11: 101915423-101915423 on Build GRCh38
rs745482845 Chr.11: 101915268-101915268 on Build GRCh38
rs771051604 Chr.11: 101915420-101915420 on Build GRCh38
rs757955230 Chr.11: 101915360-101915361 on Build GRCh38
rs1044020803 Chr.11: 101915298-101915298 on Build GRCh38
rs1007894009 Chr.11: 101915418-101915418 on Build GRCh38
rs761139539 Chr.11: 101915435-101915435 on Build GRCh38
rs190513535 Chr.11: 101915255-101915255 on Build GRCh38
rs557257305 Chr.11: 101915443-101915443 on Build GRCh38
rs777711313 Chr.11: 101915410-101915410 on Build GRCh38
rs117883580 Chr.11: 101915283-101915283 on Build GRCh38
rs748433566 Chr.11: 101915301-101915301 on Build GRCh38
rs1047431691 Chr.11: 101915293-101915293 on Build GRCh38
rs753263798 Chr.11: 101915259-101915259 on Build GRCh38
rs368939235 Chr.11: 101915265-101915265 on Build GRCh38
rs928723371 Chr.11: 101915267-101915267 on Build GRCh38
rs748048053 Chr.11: 101915366-101915366 on Build GRCh38
rs746685985 Chr.11: 101915344-101915344 on Build GRCh38
rs750002677 Chr.11: 101915293-101915293 on Build GRCh38
rs759863209 Chr.11: 101915426-101915426 on Build GRCh38
rs754752744 Chr.11: 101915349-101915349 on Build GRCh38
rs145606538 Chr.11: 101915297-101915297 on Build GRCh38
rs771824168 Chr.11: 101915436-101915436 on Build GRCh38
rs764814199 Chr.11: 101915321-101915321 on Build GRCh38
rs776043497 Chr.11: 101915431-101915431 on Build GRCh38

View Full Product Details