Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75724657-75724886 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB4
Entrez Gene ID
3691
Gene Name
integrin subunit beta 4
Gene Aliases
CD104, GP150
UniGene
Hs.632226
Gene Chromosome Location
Chr.17: 75721328-75757819 on Build GRCh38
Transcripts
NM_000213.4 NM_001005619.1 NM_001005731.2 NM_001321123.1 XM_005257309.2 XM_005257311.4 XM_006721866.3 XM_006721867.3 XM_006721868.3 XM_006721870.3 XM_011524751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4693385 c.7G>A p.G3R Chr.17: 75724710-75724710 on Build GRCh38
6306091 c.79+3G>T p.? Chr.17: 75724785-75724785 on Build GRCh38
1710789 c.3G>A p.M1I Chr.17: 75724706-75724706 on Build GRCh38
5619626 c.29C>A p.A10D Chr.17: 75724732-75724732 on Build GRCh38
3522240 c.14G>A p.R5H Chr.17: 75724717-75724717 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs113390459 Chr.17: 75724783-75724783 on Build GRCh38
rs142907560 Chr.17: 75724710-75724710 on Build GRCh38
rs202079839 Chr.17: 75724679-75724679 on Build GRCh38
rs751173051 Chr.17: 75724761-75724761 on Build GRCh38
rs747230853 Chr.17: 75724719-75724719 on Build GRCh38
rs761690140 Chr.17: 75724775-75724775 on Build GRCh38
rs756011701 Chr.17: 75724699-75724699 on Build GRCh38
rs560574772 Chr.17: 75724746-75724746 on Build GRCh38
rs765798295 Chr.17: 75724747-75724747 on Build GRCh38
rs754607957 Chr.17: 75724788-75724788 on Build GRCh38
rs368149213 Chr.17: 75724724-75724724 on Build GRCh38
rs372396780 Chr.17: 75724802-75724802 on Build GRCh38
rs780596600 Chr.17: 75724684-75724684 on Build GRCh38
rs820167 Chr.17: 75724820-75724820 on Build GRCh38
rs763352982 Chr.17: 75724752-75724752 on Build GRCh38
rs947482911 Chr.17: 75724762-75724762 on Build GRCh38
rs145773550 Chr.17: 75724760-75724760 on Build GRCh38
rs893157925 Chr.17: 75724772-75724772 on Build GRCh38
rs567076241 Chr.17: 75724713-75724713 on Build GRCh38
rs777907200 Chr.17: 75724811-75724811 on Build GRCh38
rs141077392 Chr.17: 75724716-75724716 on Build GRCh38
rs1017528017 Chr.17: 75724861-75724861 on Build GRCh38
rs375646031 Chr.17: 75724864-75724864 on Build GRCh38
rs901718611 Chr.17: 75724862-75724862 on Build GRCh38
rs766986697 Chr.17: 75724792-75724792 on Build GRCh38
rs768734852 Chr.17: 75724721-75724721 on Build GRCh38
rs147285287 Chr.17: 75724714-75724714 on Build GRCh38
rs765367145 Chr.17: 75724853-75724853 on Build GRCh38
rs934665221 Chr.17: 75724737-75724737 on Build GRCh38
rs773548728 Chr.17: 75724750-75724750 on Build GRCh38
rs748068003 Chr.17: 75724691-75724691 on Build GRCh38
rs752217597 Chr.17: 75724793-75724793 on Build GRCh38
rs753859318 Chr.17: 75724826-75724826 on Build GRCh38
rs766461494 Chr.17: 75724817-75724817 on Build GRCh38
rs374855840 Chr.17: 75724717-75724717 on Build GRCh38
rs775898450 Chr.17: 75724718-75724718 on Build GRCh38

View Full Product Details