Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 69109936-69110163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HAS3
Entrez Gene ID
3038
Gene Name
hyaluronan synthase 3
Gene Aliases
-
UniGene
Hs.592069
Gene Chromosome Location
Chr.16: 69105564-69118719 on Build GRCh38
Transcripts
NM_001199280.1 NM_005329.2 NM_138612.2 XM_005255921.2 XM_011523061.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3272045 c.582C>T p.R194R Chr.16: 69109977-69109977 on Build GRCh38
1563539 c.631A>G p.I211V Chr.16: 69110026-69110026 on Build GRCh38
3511277 c.564G>A p.Q188Q Chr.16: 69109959-69109959 on Build GRCh38
6230080 c.620C>T p.S207L Chr.16: 69110015-69110015 on Build GRCh38
3272044 c.582C>T p.R194R Chr.16: 69109977-69109977 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs762244905 Chr.16: 69110001-69110001 on Build GRCh38
rs755192495 Chr.16: 69110026-69110026 on Build GRCh38
rs898247842 Chr.16: 69110013-69110013 on Build GRCh38
rs772769711 Chr.16: 69110073-69110073 on Build GRCh38
rs371179211 Chr.16: 69110008-69110008 on Build GRCh38
rs761316634 Chr.16: 69110072-69110072 on Build GRCh38
rs370893808 Chr.16: 69110062-69110062 on Build GRCh38
rs896758135 Chr.16: 69109976-69109976 on Build GRCh38
rs371425970 Chr.16: 69110067-69110067 on Build GRCh38
rs773867261 Chr.16: 69109998-69109998 on Build GRCh38
rs745718383 Chr.16: 69110058-69110058 on Build GRCh38
rs965561479 Chr.16: 69110065-69110065 on Build GRCh38
rs768007153 Chr.16: 69110007-69110007 on Build GRCh38
rs200119038 Chr.16: 69109977-69109977 on Build GRCh38
rs995292615 Chr.16: 69110015-69110015 on Build GRCh38
rs754085372 Chr.16: 69110018-69110018 on Build GRCh38
rs1023815287 Chr.16: 69110011-69110011 on Build GRCh38
rs200883447 Chr.16: 69109989-69109989 on Build GRCh38
rs958721199 Chr.16: 69110097-69110097 on Build GRCh38
rs141994861 Chr.16: 69109992-69109992 on Build GRCh38
rs766759913 Chr.16: 69110016-69110016 on Build GRCh38
rs200276862 Chr.16: 69110039-69110039 on Build GRCh38
rs745597687 Chr.16: 69109962-69109962 on Build GRCh38
rs773835534 Chr.16: 69110078-69110078 on Build GRCh38
rs762572957 Chr.16: 69109990-69109990 on Build GRCh38
rs185816362 Chr.16: 69110082-69110082 on Build GRCh38
rs1006861823 Chr.16: 69110040-69110040 on Build GRCh38
rs539900622 Chr.16: 69110116-69110116 on Build GRCh38
rs909604977 Chr.16: 69110091-69110092 on Build GRCh38
rs757148139 Chr.16: 69110045-69110045 on Build GRCh38
rs756536611 Chr.16: 69110010-69110010 on Build GRCh38
rs375763562 Chr.16: 69110034-69110034 on Build GRCh38
rs781104620 Chr.16: 69110054-69110054 on Build GRCh38
rs773743925 Chr.16: 69110071-69110071 on Build GRCh38
rs768301385 Chr.16: 69110069-69110069 on Build GRCh38
rs914445546 Chr.16: 69110107-69110107 on Build GRCh38
rs752906964 Chr.16: 69110106-69110106 on Build GRCh38
rs779720352 Chr.16: 69110063-69110063 on Build GRCh38
rs1056687956 Chr.16: 69109961-69109961 on Build GRCh38
rs748923433 Chr.16: 69110068-69110068 on Build GRCh38
rs572521365 Chr.16: 69110088-69110088 on Build GRCh38
rs767590662 Chr.16: 69110096-69110096 on Build GRCh38
rs267604606 Chr.16: 69110031-69110031 on Build GRCh38
rs775109377 Chr.16: 69109983-69109983 on Build GRCh38
rs984264425 Chr.16: 69110090-69110090 on Build GRCh38

View Full Product Details