Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 153746936-153747207 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS3
Entrez Gene ID
65123
Gene Name
integrator complex subunit 3
Gene Aliases
C1orf193, C1orf60, INT3, SOSS-A, SOSSA
UniGene
Hs.438723
Gene Chromosome Location
Chr.1: 153728050-153774808 on Build GRCh38
Transcripts
NM_001324475.1 NM_023015.4 XM_005245459.2 XM_006711490.3 XM_011509906.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
458578 c.319-7C>T p.? Chr.1: 153746950-153746950 on Build GRCh38
896396 c.425G>A p.R142H Chr.1: 153747063-153747063 on Build GRCh38
7091175 c.432+2T>C p.? Chr.1: 153747072-153747072 on Build GRCh38
1730513 c.385A>G p.I129V Chr.1: 153747023-153747023 on Build GRCh38
3369298 c.431A>G p.Q144R Chr.1: 153747069-153747069 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145297236 Chr.1: 153747004-153747004 on Build GRCh38
rs767043117 Chr.1: 153747030-153747030 on Build GRCh38
rs753954576 Chr.1: 153747063-153747063 on Build GRCh38
rs1025337292 Chr.1: 153747167-153747167 on Build GRCh38
rs780205429 Chr.1: 153747109-153747117 on Build GRCh38
rs1009479171 Chr.1: 153747172-153747172 on Build GRCh38
rs201889970 Chr.1: 153747056-153747056 on Build GRCh38
rs956023094 Chr.1: 153747018-153747018 on Build GRCh38
rs759904170 Chr.1: 153747037-153747037 on Build GRCh38
rs140718184 Chr.1: 153747025-153747025 on Build GRCh38
rs41302547 Chr.1: 153747169-153747169 on Build GRCh38
rs772925471 Chr.1: 153747016-153747016 on Build GRCh38
rs1038013878 Chr.1: 153747129-153747129 on Build GRCh38
rs993903944 Chr.1: 153747141-153747141 on Build GRCh38
rs767775906 Chr.1: 153747039-153747039 on Build GRCh38
rs140852935 Chr.1: 153747061-153747061 on Build GRCh38
rs746928068 Chr.1: 153747119-153747119 on Build GRCh38
rs758305692 Chr.1: 153747094-153747094 on Build GRCh38
rs758224078 Chr.1: 153747106-153747107 on Build GRCh38
rs530173247 Chr.1: 153747170-153747170 on Build GRCh38
rs927097934 Chr.1: 153747097-153747097 on Build GRCh38
rs530226000 Chr.1: 153747140-153747140 on Build GRCh38
rs758696822 Chr.1: 153747075-153747075 on Build GRCh38
rs1019077430 Chr.1: 153747177-153747177 on Build GRCh38
rs779930548 Chr.1: 153747111-153747111 on Build GRCh38
rs374719274 Chr.1: 153747143-153747143 on Build GRCh38
rs544306051 Chr.1: 153747132-153747132 on Build GRCh38
rs937225726 Chr.1: 153747098-153747098 on Build GRCh38
rs776285492 Chr.1: 153746963-153746963 on Build GRCh38
rs756345533 Chr.1: 153747057-153747057 on Build GRCh38
rs774744292 Chr.1: 153747031-153747031 on Build GRCh38
rs769418464 Chr.1: 153746985-153746985 on Build GRCh38
rs747771840 Chr.1: 153746975-153746975 on Build GRCh38

View Full Product Details