Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 57752311-57752574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KATNB1
Entrez Gene ID
10300
Gene Name
katanin regulatory subunit B1
Gene Aliases
KAT, LIS6
UniGene
Hs.275675
Gene Chromosome Location
Chr.16: 57735717-57757250 on Build GRCh38
Transcripts
NM_005886.2 XM_005255772.4 XM_006721121.3 XM_006721122.2 XM_006721123.3 XM_011522810.2 XM_017022860.1 XM_017022861.1 XM_017022862.1 XM_017022863.1 XM_017022864.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6464420 c.654G>A p.L218L Chr.16: 57752551-57752551 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs782511179 Chr.16: 57752527-57752527 on Build GRCh38
rs782446656 Chr.16: 57752552-57752552 on Build GRCh38
rs782393672 Chr.16: 57752501-57752501 on Build GRCh38
rs781941098 Chr.16: 57752505-57752505 on Build GRCh38
rs782163028 Chr.16: 57752504-57752504 on Build GRCh38
rs782135998 Chr.16: 57752538-57752538 on Build GRCh38
rs540461201 Chr.16: 57752475-57752475 on Build GRCh38
rs12449078 Chr.16: 57752338-57752338 on Build GRCh38
rs782238145 Chr.16: 57752498-57752498 on Build GRCh38
rs375814930 Chr.16: 57752537-57752537 on Build GRCh38
rs7185975 Chr.16: 57752503-57752503 on Build GRCh38
rs200953449 Chr.16: 57752526-57752526 on Build GRCh38
rs1037619371 Chr.16: 57752364-57752364 on Build GRCh38
rs782804853 Chr.16: 57752463-57752463 on Build GRCh38
rs188281175 Chr.16: 57752551-57752551 on Build GRCh38
rs564194649 Chr.16: 57752412-57752412 on Build GRCh38
rs781915206 Chr.16: 57752363-57752363 on Build GRCh38
rs185034854 Chr.16: 57752496-57752496 on Build GRCh38
rs1012215683 Chr.16: 57752407-57752407 on Build GRCh38
rs568152547 Chr.16: 57752361-57752361 on Build GRCh38
rs1031610067 Chr.16: 57752478-57752478 on Build GRCh38
rs140613125 Chr.16: 57752333-57752334 on Build GRCh38
rs895158671 Chr.16: 57752368-57752368 on Build GRCh38
rs12447041 Chr.16: 57752454-57752454 on Build GRCh38
rs575274685 Chr.16: 57752328-57752328 on Build GRCh38

View Full Product Details