Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48258053-48258310 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITM2B
Entrez Gene ID
9445
Gene Name
integral membrane protein 2B
Gene Aliases
ABRI, BRI, BRI2, BRICD2B, E25B, E3-16, FBD, RDGCA, imBRI2
UniGene
Hs.643683
Gene Chromosome Location
Chr.13: 48233138-48262096 on Build GRCh38
Transcripts
NM_021999.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4148890 c.509A>T p.N170I Chr.13: 48258181-48258181 on Build GRCh38
4922293 c.522T>C p.V174V Chr.13: 48258194-48258194 on Build GRCh38
1659631 c.457C>A p.L153I Chr.13: 48258129-48258129 on Build GRCh38
6877293 c.484G>A p.D162N Chr.13: 48258156-48258156 on Build GRCh38
4943250 c.498G>T p.V166V Chr.13: 48258170-48258170 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs141232990 Chr.13: 48258172-48258172 on Build GRCh38
rs754123405 Chr.13: 48258115-48258115 on Build GRCh38
rs878981883 Chr.13: 48258120-48258120 on Build GRCh38
rs773840530 Chr.13: 48258085-48258085 on Build GRCh38
rs775340349 Chr.13: 48258282-48258282 on Build GRCh38
rs772576282 Chr.13: 48258084-48258084 on Build GRCh38
rs759922931 Chr.13: 48258272-48258272 on Build GRCh38
rs180832133 Chr.13: 48258204-48258204 on Build GRCh38
rs758048491 Chr.13: 48258189-48258189 on Build GRCh38
rs766819008 Chr.13: 48258105-48258105 on Build GRCh38
rs958229465 Chr.13: 48258203-48258203 on Build GRCh38
rs951878015 Chr.13: 48258166-48258166 on Build GRCh38
rs375626333 Chr.13: 48258279-48258279 on Build GRCh38
rs974404792 Chr.13: 48258209-48258209 on Build GRCh38
rs879231702 Chr.13: 48258123-48258123 on Build GRCh38
rs11556900 Chr.13: 48258164-48258164 on Build GRCh38
rs777134037 Chr.13: 48258191-48258191 on Build GRCh38
rs761285882 Chr.13: 48258254-48258254 on Build GRCh38
rs780778566 Chr.13: 48258089-48258090 on Build GRCh38
rs770108628 Chr.13: 48258131-48258132 on Build GRCh38
rs146916218 Chr.13: 48258275-48258275 on Build GRCh38
rs777075939 Chr.13: 48258267-48258267 on Build GRCh38
rs768875245 Chr.13: 48258208-48258208 on Build GRCh38
rs528537212 Chr.13: 48258265-48258265 on Build GRCh38
rs370730140 Chr.13: 48258250-48258250 on Build GRCh38
rs878861459 Chr.13: 48258121-48258121 on Build GRCh38
rs764934794 Chr.13: 48258177-48258177 on Build GRCh38
rs779036982 Chr.13: 48258221-48258221 on Build GRCh38
rs866835314 Chr.13: 48258153-48258153 on Build GRCh38
rs752270142 Chr.13: 48258183-48258183 on Build GRCh38
rs757109548 Chr.13: 48258198-48258198 on Build GRCh38
rs747826043 Chr.13: 48258121-48258121 on Build GRCh38
rs748368864 Chr.13: 48258229-48258229 on Build GRCh38
rs142488680 Chr.13: 48258089-48258089 on Build GRCh38
rs772199465 Chr.13: 48258240-48258240 on Build GRCh38
rs146286944 Chr.13: 48258192-48258192 on Build GRCh38

View Full Product Details