Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45005937-45006162 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HECTD3
Entrez Gene ID
79654
Gene Name
HECT domain E3 ubiquitin protein ligase 3
Gene Aliases
-
UniGene
Hs.525084
Gene Chromosome Location
Chr.1: 45002548-45011355 on Build GRCh38
Transcripts
NM_024602.5 XM_011542140.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5846828 c.910C>T p.P304S Chr.1: 45006080-45006080 on Build GRCh38
6997662 c.896G>A p.R299Q Chr.1: 45006094-45006094 on Build GRCh38
4473322 c.1842C>T p.F614F Chr.1: 45006000-45006000 on Build GRCh38
4689909 c.633C>T p.I211I Chr.1: 45006039-45006039 on Build GRCh38
6678735 c.634G>A p.G212R Chr.1: 45006038-45006038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779839902 Chr.1: 45006131-45006131 on Build GRCh38
rs758878464 Chr.1: 45005972-45005972 on Build GRCh38
rs775383501 Chr.1: 45006014-45006014 on Build GRCh38
rs778664311 Chr.1: 45006016-45006017 on Build GRCh38
rs527602464 Chr.1: 45005973-45005973 on Build GRCh38
rs749187064 Chr.1: 45006022-45006022 on Build GRCh38
rs777746576 Chr.1: 45006106-45006106 on Build GRCh38
rs770953716 Chr.1: 45006027-45006027 on Build GRCh38
rs370490198 Chr.1: 45006056-45006056 on Build GRCh38
rs769822423 Chr.1: 45006052-45006052 on Build GRCh38
rs756297767 Chr.1: 45006098-45006098 on Build GRCh38
rs372479832 Chr.1: 45005987-45005987 on Build GRCh38
rs775261801 Chr.1: 45005965-45005965 on Build GRCh38
rs760142112 Chr.1: 45005968-45005968 on Build GRCh38
rs774222736 Chr.1: 45005956-45005956 on Build GRCh38
rs759294723 Chr.1: 45005960-45005960 on Build GRCh38
rs763525446 Chr.1: 45006069-45006069 on Build GRCh38
rs756210355 Chr.1: 45006003-45006003 on Build GRCh38
rs755213463 Chr.1: 45005976-45005976 on Build GRCh38
rs1028545593 Chr.1: 45005977-45005977 on Build GRCh38
rs755405492 Chr.1: 45006078-45006078 on Build GRCh38
rs753907335 Chr.1: 45006107-45006107 on Build GRCh38
rs767858608 Chr.1: 45006082-45006082 on Build GRCh38
rs766819824 Chr.1: 45006076-45006076 on Build GRCh38
rs774311737 Chr.1: 45006038-45006038 on Build GRCh38
rs374871342 Chr.1: 45006084-45006084 on Build GRCh38
rs113135392 Chr.1: 45006120-45006120 on Build GRCh38
rs752036723 Chr.1: 45006077-45006077 on Build GRCh38
rs746642019 Chr.1: 45006133-45006133 on Build GRCh38
rs560974517 Chr.1: 45006015-45006015 on Build GRCh38
rs767231247 Chr.1: 45005961-45005961 on Build GRCh38
rs760361763 Chr.1: 45006064-45006064 on Build GRCh38
rs750949409 Chr.1: 45005969-45005969 on Build GRCh38
rs202181976 Chr.1: 45006108-45006108 on Build GRCh38
rs763396901 Chr.1: 45006072-45006072 on Build GRCh38
rs368241274 Chr.1: 45006109-45006109 on Build GRCh38
rs149994327 Chr.1: 45006112-45006112 on Build GRCh38
rs377078552 Chr.1: 45006051-45006051 on Build GRCh38
rs776136343 Chr.1: 45006071-45006071 on Build GRCh38
rs745709328 Chr.1: 45006039-45006039 on Build GRCh38

View Full Product Details