Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 43957004-43957243 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IPO13
Entrez Gene ID
9670
Gene Name
importin 13
Gene Aliases
IMP13, KAP13, LGL2, RANBP13
UniGene
Hs.158497
Gene Chromosome Location
Chr.1: 43946806-43968022 on Build GRCh38
Transcripts
NM_014652.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1321045 c.1289C>T p.T430M Chr.1: 43957212-43957212 on Build GRCh38
5473549 c.1312T>C p.L438L Chr.1: 43957235-43957235 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs1032603854 Chr.1: 43957050-43957050 on Build GRCh38
rs997719549 Chr.1: 43957059-43957059 on Build GRCh38
rs540395103 Chr.1: 43957114-43957114 on Build GRCh38
rs137908194 Chr.1: 43957128-43957128 on Build GRCh38
rs757629050 Chr.1: 43957147-43957147 on Build GRCh38
rs747888702 Chr.1: 43957218-43957218 on Build GRCh38
rs752398306 Chr.1: 43957212-43957212 on Build GRCh38
rs758394973 Chr.1: 43957177-43957177 on Build GRCh38
rs999896351 Chr.1: 43957042-43957042 on Build GRCh38
rs746166779 Chr.1: 43957175-43957175 on Build GRCh38
rs1024107441 Chr.1: 43957117-43957117 on Build GRCh38
rs756540363 Chr.1: 43957111-43957111 on Build GRCh38
rs980326002 Chr.1: 43957141-43957141 on Build GRCh38
rs904099315 Chr.1: 43957030-43957030 on Build GRCh38
rs955795642 Chr.1: 43957051-43957051 on Build GRCh38
rs531823125 Chr.1: 43957211-43957211 on Build GRCh38
rs557741009 Chr.1: 43957058-43957058 on Build GRCh38
rs780368488 Chr.1: 43957116-43957116 on Build GRCh38
rs578012350 Chr.1: 43957112-43957112 on Build GRCh38
rs776724988 Chr.1: 43957216-43957216 on Build GRCh38
rs573742411 Chr.1: 43957160-43957160 on Build GRCh38
rs369292816 Chr.1: 43957182-43957182 on Build GRCh38
rs956746817 Chr.1: 43957196-43957196 on Build GRCh38

View Full Product Details