Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 119048161-119048404 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HYOU1
Entrez Gene ID
10525
Gene Name
hypoxia up-regulated 1
Gene Aliases
GRP-170, Grp170, HSP12A, ORP-150, ORP150
UniGene
Hs.277704
Gene Chromosome Location
Chr.11: 119044185-119057246 on Build GRCh38
Transcripts
NM_001130991.2 NM_006389.4 XM_005271392.3 XM_005271393.3 XM_005271394.3 XM_017017095.1 XM_017017096.1 XM_017017097.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6537534 c.2310G>C p.E770D Chr.11: 119048314-119048314 on Build GRCh38
5987675 c.2347G>C p.E783Q Chr.11: 119048277-119048277 on Build GRCh38
2151319 c.2287delT p.S763fs*30 Chr.11: 119048337-119048337 on Build GRCh38
6519639 c.2374G>T p.V792L Chr.11: 119048250-119048250 on Build GRCh38
6367936 c.2314T>C p.S772P Chr.11: 119048310-119048310 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs782222584 Chr.11: 119048350-119048350 on Build GRCh38
rs781828400 Chr.11: 119048305-119048305 on Build GRCh38
rs782684393 Chr.11: 119048322-119048322 on Build GRCh38
rs202152542 Chr.11: 119048293-119048293 on Build GRCh38
rs781930347 Chr.11: 119048353-119048353 on Build GRCh38
rs782152879 Chr.11: 119048252-119048252 on Build GRCh38
rs536602602 Chr.11: 119048225-119048225 on Build GRCh38
rs370754185 Chr.11: 119048364-119048364 on Build GRCh38
rs782288907 Chr.11: 119048203-119048203 on Build GRCh38
rs782660545 Chr.11: 119048200-119048200 on Build GRCh38
rs782392254 Chr.11: 119048326-119048326 on Build GRCh38
rs782464197 Chr.11: 119048221-119048221 on Build GRCh38
rs890372743 Chr.11: 119048209-119048209 on Build GRCh38
rs781836147 Chr.11: 119048290-119048290 on Build GRCh38
rs144328288 Chr.11: 119048321-119048321 on Build GRCh38
rs951255131 Chr.11: 119048229-119048229 on Build GRCh38
rs782502902 Chr.11: 119048307-119048307 on Build GRCh38
rs996948821 Chr.11: 119048255-119048255 on Build GRCh38
rs924915056 Chr.11: 119048314-119048316 on Build GRCh38
rs782139288 Chr.11: 119048354-119048354 on Build GRCh38
rs377402188 Chr.11: 119048338-119048338 on Build GRCh38
rs782654390 Chr.11: 119048292-119048292 on Build GRCh38
rs149598298 Chr.11: 119048269-119048269 on Build GRCh38
rs532338554 Chr.11: 119048196-119048196 on Build GRCh38
rs782753634 Chr.11: 119048382-119048382 on Build GRCh38
rs11537773 Chr.11: 119048266-119048266 on Build GRCh38
rs550841959 Chr.11: 119048216-119048216 on Build GRCh38
rs782230111 Chr.11: 119048220-119048220 on Build GRCh38
rs376415874 Chr.11: 119048275-119048275 on Build GRCh38
rs782810900 Chr.11: 119048377-119048378 on Build GRCh38
rs113886063 Chr.11: 119048295-119048295 on Build GRCh38
rs781543896 Chr.11: 119048236-119048236 on Build GRCh38
rs11537775 Chr.11: 119048333-119048333 on Build GRCh38
rs782249856 Chr.11: 119048323-119048323 on Build GRCh38
rs782041797 Chr.11: 119048291-119048291 on Build GRCh38
rs782805574 Chr.11: 119048359-119048359 on Build GRCh38
rs782565626 Chr.11: 119048309-119048309 on Build GRCh38
rs782550453 Chr.11: 119048373-119048373 on Build GRCh38
rs992983609 Chr.11: 119048361-119048361 on Build GRCh38
rs140868450 Chr.11: 119048296-119048296 on Build GRCh38
rs569201477 Chr.11: 119048224-119048224 on Build GRCh38
rs373557923 Chr.11: 119048381-119048381 on Build GRCh38
rs545097084 Chr.11: 119048256-119048256 on Build GRCh38
rs782615631 Chr.11: 119048217-119048217 on Build GRCh38
rs782424447 Chr.11: 119048253-119048253 on Build GRCh38
rs987847949 Chr.11: 119048370-119048370 on Build GRCh38
rs782174648 Chr.11: 119048185-119048185 on Build GRCh38
rs782303315 Chr.11: 119048352-119048352 on Build GRCh38
rs781989416 Chr.11: 119048257-119048257 on Build GRCh38
rs373752157 Chr.11: 119048314-119048314 on Build GRCh38

View Full Product Details