Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 94317686-94317935 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HPGDS
Entrez Gene ID
27306
Gene Name
hematopoietic prostaglandin D synthase
Gene Aliases
GSTS, GSTS1, GSTS1-1, PGD2, PGDS
UniGene
Hs.128433
Gene Chromosome Location
Chr.4: 94298556-94342876 on Build GRCh38
Transcripts
NM_014485.2 XM_005262932.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4747640 c.221delA p.N74fs*38 Chr.4: 94317878-94317878 on Build GRCh38
2988902 c.174A>T p.G58G Chr.4: 94317925-94317925 on Build GRCh38
5401839 c.168T>A p.V56V Chr.4: 94317931-94317931 on Build GRCh38
5995714 c.181C>A p.L61I Chr.4: 94317918-94317918 on Build GRCh38
1486177 c.191G>T p.S64I Chr.4: 94317908-94317908 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs751959651 Chr.4: 94317870-94317870 on Build GRCh38
rs377012692 Chr.4: 94317859-94317859 on Build GRCh38
rs372304709 Chr.4: 94317855-94317855 on Build GRCh38
rs202112221 Chr.4: 94317864-94317864 on Build GRCh38
rs771731846 Chr.4: 94317814-94317814 on Build GRCh38
rs55672062 Chr.4: 94317860-94317860 on Build GRCh38
rs749132830 Chr.4: 94317832-94317832 on Build GRCh38
rs748002880 Chr.4: 94317854-94317854 on Build GRCh38
rs191491304 Chr.4: 94317762-94317762 on Build GRCh38
rs868082727 Chr.4: 94317872-94317872 on Build GRCh38
rs768002438 Chr.4: 94317889-94317889 on Build GRCh38
rs557447515 Chr.4: 94317862-94317862 on Build GRCh38
rs78232926 Chr.4: 94317755-94317755 on Build GRCh38
rs573793346 Chr.4: 94317790-94317790 on Build GRCh38
rs376904750 Chr.4: 94317866-94317866 on Build GRCh38
rs780214932 Chr.4: 94317831-94317831 on Build GRCh38
rs985268204 Chr.4: 94317809-94317809 on Build GRCh38
rs368980985 Chr.4: 94317891-94317891 on Build GRCh38
rs756082171 Chr.4: 94317828-94317828 on Build GRCh38
rs764765331 Chr.4: 94317867-94317867 on Build GRCh38
rs778646606 Chr.4: 94317853-94317853 on Build GRCh38
rs762210757 Chr.4: 94317875-94317875 on Build GRCh38
rs369939439 Chr.4: 94317757-94317757 on Build GRCh38
rs768627788 Chr.4: 94317833-94317833 on Build GRCh38

View Full Product Details