Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.11: 64299315-64299564 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KCNK4
Entrez Gene ID
50801
Gene Name
potassium two pore domain channel subfamily K member 4
Gene Aliases
K2p4.1, TRAAK, TRAAK1
UniGene
Hs.647233
Gene Chromosome Location
Chr.11: 64291302-64300031 on Build GRCh38
Transcripts
NM_001317090.1 NM_033310.2 NR_133660.1 NR_133661.1 NR_133662.1

Gene Symbol
KCNK4-TEX40
Entrez Gene ID
106780802
Gene Name
KCNK4-TEX40 readthrough
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.11: 64291722-64304769 on Build GRCh38
Transcripts
NM_001317090.1 NM_033310.2 NR_133660.1 NR_133661.1 NR_133662.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6425100 c.865C>T p.R289* Chr.11: 64299409-64299409 on Build GRCh38
7255650 c.990C>T p.S330S Chr.11: 64299534-64299534 on Build GRCh38
7137593 c.996C>G p.P332P Chr.11: 64299540-64299540 on Build GRCh38
4035106 c.944G>A p.G315D Chr.11: 64299488-64299488 on Build GRCh38
237593 c.1017T>C p.Y339Y Chr.11: 64299561-64299561 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761139406 Chr.11: 64299350-64299350 on Build GRCh38
rs1044774283 Chr.11: 64299410-64299410 on Build GRCh38
rs746256601 Chr.11: 64299534-64299534 on Build GRCh38
rs751508722 Chr.11: 64299383-64299383 on Build GRCh38
rs953778 Chr.11: 64299527-64299527 on Build GRCh38
rs768358494 Chr.11: 64299470-64299470 on Build GRCh38
rs775313901 Chr.11: 64299360-64299360 on Build GRCh38
rs755091552 Chr.11: 64299509-64299509 on Build GRCh38
rs763982687 Chr.11: 64299513-64299513 on Build GRCh38
rs754106221 Chr.11: 64299424-64299424 on Build GRCh38
rs1054094323 Chr.11: 64299503-64299503 on Build GRCh38
rs775262935 Chr.11: 64299468-64299468 on Build GRCh38
rs775736273 Chr.11: 64299507-64299508 on Build GRCh38
rs556442938 Chr.11: 64299432-64299432 on Build GRCh38
rs762521639 Chr.11: 64299379-64299379 on Build GRCh38
rs748268121 Chr.11: 64299435-64299435 on Build GRCh38
rs747339416 Chr.11: 64299445-64299445 on Build GRCh38
rs867027412 Chr.11: 64299526-64299526 on Build GRCh38
rs904988443 Chr.11: 64299411-64299411 on Build GRCh38
rs933141425 Chr.11: 64299408-64299408 on Build GRCh38
rs1030678468 Chr.11: 64299521-64299521 on Build GRCh38
rs201358336 Chr.11: 64299517-64299517 on Build GRCh38
rs773676493 Chr.11: 64299481-64299481 on Build GRCh38
rs768144352 Chr.11: 64299536-64299536 on Build GRCh38
rs760437142 Chr.11: 64299497-64299497 on Build GRCh38
rs2232417 Chr.11: 64299387-64299387 on Build GRCh38
rs752675226 Chr.11: 64299512-64299512 on Build GRCh38
rs778367342 Chr.11: 64299438-64299438 on Build GRCh38
rs746327186 Chr.11: 64299370-64299371 on Build GRCh38
rs758778655 Chr.11: 64299436-64299436 on Build GRCh38
rs772483824 Chr.11: 64299340-64299340 on Build GRCh38
rs747576337 Chr.11: 64299543-64299543 on Build GRCh38
rs993406941 Chr.11: 64299515-64299515 on Build GRCh38
rs771658589 Chr.11: 64299545-64299545 on Build GRCh38
rs765512931 Chr.11: 64299510-64299510 on Build GRCh38
rs757684837 Chr.11: 64299523-64299523 on Build GRCh38
rs770293176 Chr.11: 64299492-64299506 on Build GRCh38
rs866996839 Chr.11: 64299349-64299349 on Build GRCh38
rs780215757 Chr.11: 64299403-64299403 on Build GRCh38
rs972153727 Chr.11: 64299381-64299381 on Build GRCh38
rs755931043 Chr.11: 64299396-64299396 on Build GRCh38
rs774084627 Chr.11: 64299537-64299537 on Build GRCh38
rs769522534 Chr.11: 64299353-64299353 on Build GRCh38
rs1050288979 Chr.11: 64299465-64299465 on Build GRCh38
rs576679008 Chr.11: 64299508-64299508 on Build GRCh38
rs773386180 Chr.11: 64299346-64299346 on Build GRCh38
rs761408425 Chr.11: 64299482-64299482 on Build GRCh38
rs763562553 Chr.11: 64299380-64299380 on Build GRCh38
rs767313879 Chr.11: 64299488-64299488 on Build GRCh38
rs747568761 Chr.11: 64299520-64299520 on Build GRCh38
rs767503396 Chr.11: 64299392-64299392 on Build GRCh38
rs755074316 Chr.11: 64299429-64299429 on Build GRCh38
rs773170113 Chr.11: 64299492-64299492 on Build GRCh38

View Full Product Details