Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 12121604-12121837 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIVEP1
Entrez Gene ID
3096
Gene Name
human immunodeficiency virus type I enhancer binding protein 1
Gene Aliases
CIRIP, CRYBP1, GAAP, MBP-1, PRDII-BF1, Schnurri-1, ZAS1, ZNF40, ZNF40A
UniGene
Hs.567284
Gene Chromosome Location
Chr.6: 12002096-12212034 on Build GRCh38
Transcripts
NM_002114.3 XM_011514546.2 XM_011514547.2 XM_011514548.2 XM_011514549.2 XM_011514550.2 XM_011514551.2 XM_011514552.2 XM_011514553.1 XM_011514555.2 XM_017010800.1 XM_017010801.1 XM_017010802.1 XM_017010803.1 XM_017010804.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4424797 c.1991G>A p.S664N Chr.6: 12121786-12121786 on Build GRCh38
5018137 c.1867_1880del14 p.Q624fs*52 Chr.6: 12121662-12121675 on Build GRCh38
1137421 c.2007A>C p.G669G Chr.6: 12121802-12121802 on Build GRCh38
3171027 c.2004A>G p.L668L Chr.6: 12121799-12121799 on Build GRCh38
483348 c.1956C>A p.T652T Chr.6: 12121751-12121751 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs765437770 Chr.6: 12121661-12121661 on Build GRCh38
rs200876372 Chr.6: 12121707-12121707 on Build GRCh38
rs746836276 Chr.6: 12121792-12121792 on Build GRCh38
rs779581742 Chr.6: 12121701-12121701 on Build GRCh38
rs771990986 Chr.6: 12121626-12121626 on Build GRCh38
rs964651822 Chr.6: 12121774-12121774 on Build GRCh38
rs550663223 Chr.6: 12121718-12121718 on Build GRCh38
rs776616094 Chr.6: 12121658-12121658 on Build GRCh38
rs747869688 Chr.6: 12121710-12121710 on Build GRCh38
rs758086842 Chr.6: 12121783-12121783 on Build GRCh38
rs757212970 Chr.6: 12121797-12121797 on Build GRCh38
rs778495637 Chr.6: 12121709-12121709 on Build GRCh38
rs746919379 Chr.6: 12121716-12121716 on Build GRCh38
rs998519563 Chr.6: 12121690-12121690 on Build GRCh38
rs746004351 Chr.6: 12121725-12121725 on Build GRCh38
rs774900283 Chr.6: 12121747-12121747 on Build GRCh38
rs548393511 Chr.6: 12121651-12121651 on Build GRCh38
rs1034428674 Chr.6: 12121662-12121662 on Build GRCh38
rs755535486 Chr.6: 12121682-12121682 on Build GRCh38
rs753335558 Chr.6: 12121704-12121704 on Build GRCh38
rs753385436 Chr.6: 12121765-12121765 on Build GRCh38
rs777521235 Chr.6: 12121791-12121791 on Build GRCh38
rs752139266 Chr.6: 12121675-12121675 on Build GRCh38
rs376299465 Chr.6: 12121780-12121780 on Build GRCh38
rs745899267 Chr.6: 12121807-12121807 on Build GRCh38
rs759783535 Chr.6: 12121660-12121660 on Build GRCh38
rs200095122 Chr.6: 12121688-12121688 on Build GRCh38
rs763356501 Chr.6: 12121735-12121735 on Build GRCh38
rs760642384 Chr.6: 12121717-12121717 on Build GRCh38
rs780993174 Chr.6: 12121806-12121806 on Build GRCh38
rs539496550 Chr.6: 12121736-12121736 on Build GRCh38
rs754452579 Chr.6: 12121768-12121768 on Build GRCh38
rs764813557 Chr.6: 12121769-12121769 on Build GRCh38
rs768313308 Chr.6: 12121775-12121775 on Build GRCh38
rs748901124 Chr.6: 12121779-12121781 on Build GRCh38
rs776760535 Chr.6: 12121724-12121724 on Build GRCh38
rs187187140 Chr.6: 12121673-12121673 on Build GRCh38
rs369522683 Chr.6: 12121705-12121705 on Build GRCh38
rs201669166 Chr.6: 12121711-12121711 on Build GRCh38
rs775871857 Chr.6: 12121730-12121730 on Build GRCh38
rs369499373 Chr.6: 12121808-12121808 on Build GRCh38
rs201768344 Chr.6: 12121674-12121674 on Build GRCh38
rs932054806 Chr.6: 12121734-12121734 on Build GRCh38
rs971130060 Chr.6: 12121634-12121634 on Build GRCh38
rs760689832 Chr.6: 12121639-12121639 on Build GRCh38
rs773137527 Chr.6: 12121632-12121632 on Build GRCh38
rs1038999430 Chr.6: 12121652-12121652 on Build GRCh38
rs762362516 Chr.6: 12121752-12121752 on Build GRCh38
rs1031425893 Chr.6: 12121702-12121702 on Build GRCh38
rs767968913 Chr.6: 12121762-12121762 on Build GRCh38
rs543109288 Chr.6: 12121666-12121666 on Build GRCh38
rs769994799 Chr.6: 12121727-12121727 on Build GRCh38

View Full Product Details