Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 113956446-113956673 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIPK1
Entrez Gene ID
204851
Gene Name
homeodomain interacting protein kinase 1
Gene Aliases
Myak, Nbak2
UniGene
Hs.532363
Gene Chromosome Location
Chr.1: 113929197-113977869 on Build GRCh38
Transcripts
NM_152696.3 NM_181358.2 NM_198268.2 NM_198269.2 XM_005270609.3 XM_005270610.4 XM_005270611.4 XM_005270612.4 XM_005270613.4 XM_006710443.3 XM_011540977.2 XM_017000605.1 XM_017000606.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4466295 c.1426C>T p.L476L Chr.1: 113956645-113956645 on Build GRCh38
1683778 c.1429delG p.E477fs*19 Chr.1: 113956648-113956648 on Build GRCh38
4965069 c.1408-4G>A p.? Chr.1: 113956623-113956623 on Build GRCh38
4965070 c.1408-4G>A p.? Chr.1: 113956623-113956623 on Build GRCh38
1683779 c.1429delG p.E477fs*19 Chr.1: 113956648-113956648 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs932930601 Chr.1: 113956512-113956512 on Build GRCh38
rs778750886 Chr.1: 113956633-113956633 on Build GRCh38
rs11546190 Chr.1: 113956641-113956641 on Build GRCh38
rs887737220 Chr.1: 113956575-113956575 on Build GRCh38
rs979100302 Chr.1: 113956485-113956485 on Build GRCh38
rs115873210 Chr.1: 113956511-113956511 on Build GRCh38
rs935105540 Chr.1: 113956535-113956535 on Build GRCh38
rs1050772239 Chr.1: 113956556-113956556 on Build GRCh38
rs1040168711 Chr.1: 113956541-113956541 on Build GRCh38
rs931891515 Chr.1: 113956546-113956546 on Build GRCh38
rs1050060087 Chr.1: 113956535-113956536 on Build GRCh38
rs201763810 Chr.1: 113956639-113956639 on Build GRCh38
rs755624283 Chr.1: 113956614-113956614 on Build GRCh38
rs570207068 Chr.1: 113956563-113956563 on Build GRCh38
rs968125995 Chr.1: 113956473-113956473 on Build GRCh38
rs41283524 Chr.1: 113956582-113956582 on Build GRCh38
rs747787897 Chr.1: 113956634-113956634 on Build GRCh38
rs767240739 Chr.1: 113956572-113956572 on Build GRCh38
rs1038405151 Chr.1: 113956601-113956601 on Build GRCh38
rs890122152 Chr.1: 113956558-113956558 on Build GRCh38
rs923109322 Chr.1: 113956542-113956542 on Build GRCh38

View Full Product Details