Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 156751164-156751431 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HDGF
Entrez Gene ID
3068
Gene Name
hepatoma-derived growth factor
Gene Aliases
HMG1L2
UniGene
Hs.743948
Gene Chromosome Location
Chr.1: 156742107-156752448 on Build GRCh38
Transcripts
NM_001126050.1 NM_001126051.1 NM_001319186.1 NM_001319187.1 NM_001319188.1 NM_004494.2 NR_135008.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3934148 c.52G>A p.A18T Chr.1: 156751378-156751378 on Build GRCh38
4548914 c.45G>C p.L15L Chr.1: 156751385-156751385 on Build GRCh38
3802249 c.87+2T>G p.? Chr.1: 156751341-156751341 on Build GRCh38
6883403 c.30C>T p.Y10Y Chr.1: 156751400-156751400 on Build GRCh38
6867344 c.57G>T p.K19N Chr.1: 156751373-156751373 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs906136553 Chr.1: 156751264-156751264 on Build GRCh38
rs763557184 Chr.1: 156751378-156751378 on Build GRCh38
rs777643812 Chr.1: 156751311-156751311 on Build GRCh38
rs889516959 Chr.1: 156751361-156751361 on Build GRCh38
rs773796622 Chr.1: 156751376-156751376 on Build GRCh38
rs759201444 Chr.1: 156751414-156751414 on Build GRCh38
rs1010613861 Chr.1: 156751296-156751296 on Build GRCh38
rs993355151 Chr.1: 156751245-156751245 on Build GRCh38
rs909855890 Chr.1: 156751286-156751286 on Build GRCh38
rs555647453 Chr.1: 156751230-156751230 on Build GRCh38
rs781301755 Chr.1: 156751335-156751335 on Build GRCh38
rs756043971 Chr.1: 156751310-156751310 on Build GRCh38
rs747242740 Chr.1: 156751319-156751319 on Build GRCh38
rs570988018 Chr.1: 156751412-156751412 on Build GRCh38
rs953749189 Chr.1: 156751197-156751197 on Build GRCh38
rs749240762 Chr.1: 156751312-156751312 on Build GRCh38
rs12030654 Chr.1: 156751240-156751240 on Build GRCh38
rs559929989 Chr.1: 156751231-156751231 on Build GRCh38
rs546207871 Chr.1: 156751260-156751260 on Build GRCh38
rs966608741 Chr.1: 156751217-156751217 on Build GRCh38
rs915994136 Chr.1: 156751270-156751270 on Build GRCh38
rs747100192 Chr.1: 156751412-156751412 on Build GRCh38
rs1037166436 Chr.1: 156751214-156751214 on Build GRCh38
rs767037808 Chr.1: 156751305-156751305 on Build GRCh38
rs748138050 Chr.1: 156751354-156751354 on Build GRCh38
rs371694112 Chr.1: 156751322-156751322 on Build GRCh38
rs531956693 Chr.1: 156751367-156751367 on Build GRCh38
rs1037247490 Chr.1: 156751307-156751307 on Build GRCh38
rs950255809 Chr.1: 156751235-156751235 on Build GRCh38
rs1022195065 Chr.1: 156751211-156751211 on Build GRCh38
rs948723464 Chr.1: 156751277-156751277 on Build GRCh38
rs768810835 Chr.1: 156751323-156751323 on Build GRCh38
rs143905023 Chr.1: 156751284-156751284 on Build GRCh38
rs1001858307 Chr.1: 156751275-156751275 on Build GRCh38
rs998347372 Chr.1: 156751190-156751190 on Build GRCh38
rs954060685 Chr.1: 156751195-156751195 on Build GRCh38
rs1028040795 Chr.1: 156751192-156751192 on Build GRCh38
rs774910787 Chr.1: 156751410-156751410 on Build GRCh38
rs771415808 Chr.1: 156751408-156751408 on Build GRCh38
rs1049508476 Chr.1: 156751336-156751336 on Build GRCh38
rs1055237386 Chr.1: 156751282-156751282 on Build GRCh38
rs940285488 Chr.1: 156751288-156751288 on Build GRCh38
rs752195032 Chr.1: 156751306-156751306 on Build GRCh38
rs1010528584 Chr.1: 156751200-156751200 on Build GRCh38
rs868163154 Chr.1: 156751393-156751393 on Build GRCh38
rs756982976 Chr.1: 156751313-156751313 on Build GRCh38
rs778748862 Chr.1: 156751318-156751318 on Build GRCh38

View Full Product Details