Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 21409335-21409574 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFNA8
Entrez Gene ID
3445
Gene Name
interferon alpha 8
Gene Aliases
IFN-alphaB
UniGene
Hs.73890
Gene Chromosome Location
Chr.9: 21409147-21410185 on Build GRCh38
Transcripts
NM_002170.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6683891 c.264C>A p.F88L Chr.9: 21409440-21409440 on Build GRCh38
4836428 c.352G>A p.D118N Chr.9: 21409528-21409528 on Build GRCh38
6683893 c.379G>A p.V127M Chr.9: 21409555-21409555 on Build GRCh38
487321 c.311T>A p.L104H Chr.9: 21409487-21409487 on Build GRCh38
4920152 c.389T>C p.I130T Chr.9: 21409565-21409565 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs749581717 Chr.9: 21409398-21409398 on Build GRCh38
rs776474533 Chr.9: 21409382-21409382 on Build GRCh38
rs147449554 Chr.9: 21409479-21409479 on Build GRCh38
rs116195490 Chr.9: 21409507-21409507 on Build GRCh38
rs199520715 Chr.9: 21409506-21409506 on Build GRCh38
rs748630324 Chr.9: 21409431-21409431 on Build GRCh38
rs776652981 Chr.9: 21409456-21409456 on Build GRCh38
rs758893587 Chr.9: 21409538-21409538 on Build GRCh38
rs766153547 Chr.9: 21409380-21409380 on Build GRCh38
rs538648951 Chr.9: 21409476-21409476 on Build GRCh38
rs779417192 Chr.9: 21409405-21409405 on Build GRCh38
rs775620362 Chr.9: 21409465-21409465 on Build GRCh38
rs1048186496 Chr.9: 21409436-21409436 on Build GRCh38
rs760920714 Chr.9: 21409379-21409379 on Build GRCh38
rs943981638 Chr.9: 21409373-21409373 on Build GRCh38
rs370545982 Chr.9: 21409461-21409461 on Build GRCh38
rs572044600 Chr.9: 21409499-21409499 on Build GRCh38
rs772089007 Chr.9: 21409370-21409370 on Build GRCh38
rs751089846 Chr.9: 21409486-21409486 on Build GRCh38
rs752644411 Chr.9: 21409391-21409391 on Build GRCh38
rs374095498 Chr.9: 21409451-21409451 on Build GRCh38
rs747117991 Chr.9: 21409552-21409552 on Build GRCh38
rs772178929 Chr.9: 21409442-21409442 on Build GRCh38
rs376876265 Chr.9: 21409535-21409535 on Build GRCh38
rs762576266 Chr.9: 21409392-21409392 on Build GRCh38
rs779215515 Chr.9: 21409531-21409531 on Build GRCh38
rs147495176 Chr.9: 21409511-21409511 on Build GRCh38
rs1016554964 Chr.9: 21409434-21409434 on Build GRCh38
rs755383090 Chr.9: 21409525-21409525 on Build GRCh38
rs778084720 Chr.9: 21409545-21409545 on Build GRCh38
rs762956897 Chr.9: 21409466-21409466 on Build GRCh38
rs868066730 Chr.9: 21409541-21409541 on Build GRCh38
rs148720237 Chr.9: 21409399-21409399 on Build GRCh38
rs910751556 Chr.9: 21409540-21409540 on Build GRCh38
rs776562303 Chr.9: 21409396-21409396 on Build GRCh38
rs763679295 Chr.9: 21409393-21409393 on Build GRCh38
rs973080391 Chr.9: 21409389-21409389 on Build GRCh38
rs751214833 Chr.9: 21409395-21409395 on Build GRCh38
rs777825664 Chr.9: 21409444-21409444 on Build GRCh38
rs200595461 Chr.9: 21409455-21409455 on Build GRCh38
rs773470949 Chr.9: 21409378-21409378 on Build GRCh38
rs371792646 Chr.9: 21409459-21409459 on Build GRCh38
rs759195852 Chr.9: 21409383-21409383 on Build GRCh38
rs768605643 Chr.9: 21409372-21409372 on Build GRCh38
rs906640565 Chr.9: 21409361-21409361 on Build GRCh38
rs765272000 Chr.9: 21409386-21409386 on Build GRCh38
rs755254991 Chr.9: 21409404-21409404 on Build GRCh38
rs749231173 Chr.9: 21409406-21409406 on Build GRCh38

View Full Product Details