Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 142942337-142942608 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KEL
Entrez Gene ID
3792
Gene Name
Kell blood group, metallo-endopeptidase
Gene Aliases
CD238, ECE3
UniGene
Hs.368588
Gene Chromosome Location
Chr.7: 142941114-142962416 on Build GRCh38
Transcripts
NM_000420.2 XM_005249993.2 XM_005249994.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5024101 c.2013G>T p.Q671H Chr.7: 142942458-142942458 on Build GRCh38
6109356 c.1986G>T p.L662L Chr.7: 142942485-142942485 on Build GRCh38
4569770 c.2015T>C p.I672T Chr.7: 142942456-142942456 on Build GRCh38
3635034 c.1955G>A p.R652K Chr.7: 142942516-142942516 on Build GRCh38
1673601 c.1976A>G p.E659G Chr.7: 142942495-142942495 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs202116062 Chr.7: 142942424-142942424 on Build GRCh38
rs752599863 Chr.7: 142942455-142942455 on Build GRCh38
rs868367052 Chr.7: 142942529-142942529 on Build GRCh38
rs751310423 Chr.7: 142942466-142942466 on Build GRCh38
rs374463633 Chr.7: 142942386-142942386 on Build GRCh38
rs918544290 Chr.7: 142942502-142942502 on Build GRCh38
rs41312165 Chr.7: 142942360-142942360 on Build GRCh38
rs61729051 Chr.7: 142942444-142942444 on Build GRCh38
rs761958724 Chr.7: 142942439-142942439 on Build GRCh38
rs760716472 Chr.7: 142942447-142942447 on Build GRCh38
rs777575613 Chr.7: 142942392-142942392 on Build GRCh38
rs368716327 Chr.7: 142942570-142942570 on Build GRCh38
rs200880305 Chr.7: 142942541-142942541 on Build GRCh38
rs370938244 Chr.7: 142942508-142942508 on Build GRCh38
rs1027204995 Chr.7: 142942417-142942417 on Build GRCh38
rs758315599 Chr.7: 142942456-142942456 on Build GRCh38
rs143804816 Chr.7: 142942359-142942359 on Build GRCh38
rs950747451 Chr.7: 142942433-142942433 on Build GRCh38
rs371862828 Chr.7: 142942414-142942414 on Build GRCh38
rs750119372 Chr.7: 142942540-142942540 on Build GRCh38
rs777434899 Chr.7: 142942457-142942457 on Build GRCh38
rs769921538 Chr.7: 142942517-142942517 on Build GRCh38
rs757027102 Chr.7: 142942467-142942467 on Build GRCh38
rs751376270 Chr.7: 142942519-142942519 on Build GRCh38
rs746672530 Chr.7: 142942402-142942402 on Build GRCh38
rs1042359399 Chr.7: 142942496-142942496 on Build GRCh38
rs770640969 Chr.7: 142942409-142942409 on Build GRCh38
rs61729047 Chr.7: 142942448-142942448 on Build GRCh38
rs768309531 Chr.7: 142942500-142942500 on Build GRCh38
rs774640107 Chr.7: 142942489-142942489 on Build GRCh38
rs371080072 Chr.7: 142942385-142942385 on Build GRCh38
rs891209890 Chr.7: 142942557-142942557 on Build GRCh38
rs748821936 Chr.7: 142942563-142942563 on Build GRCh38
rs566187145 Chr.7: 142942561-142942561 on Build GRCh38
rs902425813 Chr.7: 142942524-142942524 on Build GRCh38
rs964970218 Chr.7: 142942397-142942397 on Build GRCh38
rs776707033 Chr.7: 142942506-142942506 on Build GRCh38
rs769403880 Chr.7: 142942480-142942480 on Build GRCh38
rs372807961 Chr.7: 142942565-142942565 on Build GRCh38
rs368090451 Chr.7: 142942492-142942492 on Build GRCh38
rs748563011 Chr.7: 142942578-142942578 on Build GRCh38
rs529389432 Chr.7: 142942516-142942516 on Build GRCh38
rs757084043 Chr.7: 142942530-142942530 on Build GRCh38
rs769151837 Chr.7: 142942488-142942488 on Build GRCh38
rs140592001 Chr.7: 142942485-142942485 on Build GRCh38
rs973601828 Chr.7: 142942423-142942423 on Build GRCh38
rs199919501 Chr.7: 142942413-142942413 on Build GRCh38
rs774715374 Chr.7: 142942432-142942432 on Build GRCh38
rs147464117 Chr.7: 142942441-142942441 on Build GRCh38
rs767325080 Chr.7: 142942538-142942538 on Build GRCh38
rs1056095842 Chr.7: 142942556-142942556 on Build GRCh38
rs773401783 Chr.7: 142942499-142942499 on Build GRCh38
rs779446787 Chr.7: 142942566-142942566 on Build GRCh38
rs374949198 Chr.7: 142942420-142942420 on Build GRCh38
rs374739410 Chr.7: 142942476-142942476 on Build GRCh38
rs768921121 Chr.7: 142942425-142942425 on Build GRCh38

View Full Product Details