Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.6: 146399447-146399716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRM1
Entrez Gene ID
2911
Gene Name
glutamate metabotropic receptor 1
Gene Aliases
GPRC1A, MGLU1, MGLUR1, PPP1R85, SCAR13
UniGene
Hs.32945
Gene Chromosome Location
Chr.6: 146027634-146437598 on Build GRCh38
Transcripts
NM_001278064.1 NM_001278065.1 NM_001278066.1 NM_001278067.1 XM_011535782.1 XM_017010783.1 XM_017010784.1 XM_017010785.1 XM_017010786.1 XM_017010787.1 XM_017010788.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1074274 c.2637G>T p.K879N Chr.6: 146399676-146399676 on Build GRCh38
6675820 c.2502G>T p.K834N Chr.6: 146399541-146399541 on Build GRCh38
3829247 c.2527G>A p.E843K Chr.6: 146399566-146399566 on Build GRCh38
2154795 c.2583C>T p.G861G Chr.6: 146399622-146399622 on Build GRCh38
740687 c.2650G>C p.G884R Chr.6: 146399689-146399689 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs761307645 Chr.6: 146399544-146399544 on Build GRCh38
rs747520531 Chr.6: 146399571-146399571 on Build GRCh38
rs376290378 Chr.6: 146399686-146399686 on Build GRCh38
rs764439606 Chr.6: 146399684-146399684 on Build GRCh38
rs362936 Chr.6: 146399690-146399690 on Build GRCh38
rs550219072 Chr.6: 146399641-146399641 on Build GRCh38
rs1050532160 Chr.6: 146399602-146399602 on Build GRCh38
rs371375072 Chr.6: 146399648-146399648 on Build GRCh38
rs211694392 Chr.6: 146399691-146399693 on Build GRCh38
rs773205376 Chr.6: 146399634-146399634 on Build GRCh38
rs778194565 Chr.6: 146399562-146399562 on Build GRCh38
rs142675468 Chr.6: 146399535-146399535 on Build GRCh38
rs372602926 Chr.6: 146399570-146399570 on Build GRCh38
rs143544032 Chr.6: 146399620-146399620 on Build GRCh38
rs765450864 Chr.6: 146399658-146399658 on Build GRCh38
rs993938006 Chr.6: 146399586-146399586 on Build GRCh38
rs770540155 Chr.6: 146399601-146399603 on Build GRCh38
rs765500811 Chr.6: 146399661-146399661 on Build GRCh38
rs35449363 Chr.6: 146399691-146399692 on Build GRCh38
rs1026299344 Chr.6: 146399616-146399616 on Build GRCh38
rs755202615 Chr.6: 146399523-146399523 on Build GRCh38
rs764746524 Chr.6: 146399576-146399576 on Build GRCh38
rs758693677 Chr.6: 146399679-146399679 on Build GRCh38
rs773909784 Chr.6: 146399625-146399625 on Build GRCh38
rs766272199 Chr.6: 146399642-146399642 on Build GRCh38
rs757829503 Chr.6: 146399592-146399592 on Build GRCh38
rs758783319 Chr.6: 146399551-146399551 on Build GRCh38
rs779602255 Chr.6: 146399610-146399610 on Build GRCh38
rs772008860 Chr.6: 146399631-146399631 on Build GRCh38
rs747905402 Chr.6: 146399629-146399629 on Build GRCh38
rs867554621 Chr.6: 146399500-146399500 on Build GRCh38
rs148042148 Chr.6: 146399622-146399622 on Build GRCh38

View Full Product Details