Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 75731149-75731410 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGB4
Entrez Gene ID
3691
Gene Name
integrin subunit beta 4
Gene Aliases
CD104, GP150
UniGene
Hs.632226
Gene Chromosome Location
Chr.17: 75721328-75757819 on Build GRCh38
Transcripts
NM_000213.4 NM_001005619.1 NM_001005731.2 NM_001321123.1 XM_005257309.2 XM_005257311.4 XM_006721866.3 XM_006721867.3 XM_006721868.3 XM_006721870.3 XM_011524751.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3522260 c.1144C>T p.R382W Chr.17: 75731297-75731297 on Build GRCh38
4693394 c.1215+2T>C p.? Chr.17: 75731370-75731370 on Build GRCh38
7164157 c.1190C>T p.S397F Chr.17: 75731343-75731343 on Build GRCh38
4069647 c.1170C>T p.F390F Chr.17: 75731323-75731323 on Build GRCh38
4693393 c.1215+2T>C p.? Chr.17: 75731370-75731370 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs748324133 Chr.17: 75731253-75731253 on Build GRCh38
rs772693450 Chr.17: 75731364-75731364 on Build GRCh38
rs867940087 Chr.17: 75731250-75731250 on Build GRCh38
rs779037959 Chr.17: 75731252-75731252 on Build GRCh38
rs748554997 Chr.17: 75731358-75731358 on Build GRCh38
rs367872059 Chr.17: 75731289-75731289 on Build GRCh38
rs201686414 Chr.17: 75731387-75731387 on Build GRCh38
rs964677566 Chr.17: 75731178-75731178 on Build GRCh38
rs754774133 Chr.17: 75731224-75731224 on Build GRCh38
rs755924283 Chr.17: 75731381-75731381 on Build GRCh38
rs141970956 Chr.17: 75731248-75731248 on Build GRCh38
rs886053409 Chr.17: 75731363-75731363 on Build GRCh38
rs144655991 Chr.17: 75731334-75731334 on Build GRCh38
rs773508913 Chr.17: 75731355-75731355 on Build GRCh38
rs973315793 Chr.17: 75731378-75731378 on Build GRCh38
rs772335175 Chr.17: 75731332-75731332 on Build GRCh38
rs780184035 Chr.17: 75731344-75731344 on Build GRCh38
rs141337579 Chr.17: 75731297-75731297 on Build GRCh38
rs772224695 Chr.17: 75731266-75731266 on Build GRCh38
rs1010473905 Chr.17: 75731241-75731241 on Build GRCh38
rs767417049 Chr.17: 75731326-75731326 on Build GRCh38
rs754973172 Chr.17: 75731321-75731321 on Build GRCh38
rs745545992 Chr.17: 75731357-75731358 on Build GRCh38
rs200122430 Chr.17: 75731354-75731354 on Build GRCh38
rs907459726 Chr.17: 75731362-75731362 on Build GRCh38
rs767613447 Chr.17: 75731373-75731373 on Build GRCh38
rs190295755 Chr.17: 75731175-75731175 on Build GRCh38
rs777045339 Chr.17: 75731288-75731288 on Build GRCh38
rs369393539 Chr.17: 75731383-75731383 on Build GRCh38
rs768938705 Chr.17: 75731273-75731273 on Build GRCh38
rs781149755 Chr.17: 75731226-75731226 on Build GRCh38
rs1031818656 Chr.17: 75731368-75731368 on Build GRCh38
rs751396834 Chr.17: 75731319-75731319 on Build GRCh38
rs761081268 Chr.17: 75731384-75731384 on Build GRCh38
rs749456201 Chr.17: 75731247-75731247 on Build GRCh38
rs892066882 Chr.17: 75731230-75731230 on Build GRCh38
rs563495764 Chr.17: 75731357-75731357 on Build GRCh38
rs374254754 Chr.17: 75731365-75731365 on Build GRCh38
rs755444323 Chr.17: 75731392-75731392 on Build GRCh38
rs552307377 Chr.17: 75731372-75731372 on Build GRCh38
rs762334700 Chr.17: 75731298-75731298 on Build GRCh38
rs143833423 Chr.17: 75731390-75731390 on Build GRCh38
rs779901316 Chr.17: 75731207-75731207 on Build GRCh38
rs775809934 Chr.17: 75731271-75731271 on Build GRCh38
rs376396837 Chr.17: 75731220-75731220 on Build GRCh38
rs373755025 Chr.17: 75731214-75731214 on Build GRCh38
rs774074057 Chr.17: 75731367-75731367 on Build GRCh38
rs886053408 Chr.17: 75731246-75731246 on Build GRCh38
rs765939074 Chr.17: 75731306-75731306 on Build GRCh38
rs1057316705 Chr.17: 75731217-75731217 on Build GRCh38
rs528068970 Chr.17: 75731389-75731389 on Build GRCh38
rs752599095 Chr.17: 75731331-75731331 on Build GRCh38

View Full Product Details