Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 92242167-92242402 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IARS
Entrez Gene ID
3376
Gene Name
isoleucyl-tRNA synthetase
Gene Aliases
IARS1, ILERS, ILRS, IRS, PRO0785
UniGene
Hs.445403
Gene Chromosome Location
Chr.9: 92210207-92293756 on Build GRCh38
Transcripts
NM_002161.5 NM_013417.3 NR_073446.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4541765 c.225G>A p.L75L Chr.9: 92242265-92242265 on Build GRCh38
3413810 c.305C>G p.S102W Chr.9: 92242185-92242185 on Build GRCh38
3413809 c.3146C>G p.S1049W Chr.9: 92242185-92242185 on Build GRCh38
3659503 c.264C>T p.T88T Chr.9: 92242226-92242226 on Build GRCh38
5979704 c.3005A>G p.N1002S Chr.9: 92242326-92242326 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs145404152 Chr.9: 92242206-92242206 on Build GRCh38
rs747247907 Chr.9: 92242325-92242325 on Build GRCh38
rs200927999 Chr.9: 92242361-92242361 on Build GRCh38
rs761410657 Chr.9: 92242365-92242365 on Build GRCh38
rs780187451 Chr.9: 92242321-92242321 on Build GRCh38
rs768728111 Chr.9: 92242213-92242213 on Build GRCh38
rs533157314 Chr.9: 92242349-92242349 on Build GRCh38
rs1043969518 Chr.9: 92242234-92242234 on Build GRCh38
rs755039797 Chr.9: 92242326-92242326 on Build GRCh38
rs373950398 Chr.9: 92242352-92242352 on Build GRCh38
rs34737051 Chr.9: 92242195-92242195 on Build GRCh38
rs763708022 Chr.9: 92242283-92242283 on Build GRCh38
rs765626247 Chr.9: 92242368-92242368 on Build GRCh38
rs770757029 Chr.9: 92242223-92242223 on Build GRCh38
rs749217090 Chr.9: 92242333-92242333 on Build GRCh38
rs765617323 Chr.9: 92242293-92242293 on Build GRCh38
rs773078041 Chr.9: 92242218-92242218 on Build GRCh38
rs758825266 Chr.9: 92242320-92242320 on Build GRCh38
rs201773352 Chr.9: 92242240-92242240 on Build GRCh38
rs779296648 Chr.9: 92242340-92242341 on Build GRCh38
rs1031102724 Chr.9: 92242275-92242275 on Build GRCh38
rs545812051 Chr.9: 92242346-92242347 on Build GRCh38
rs201431564 Chr.9: 92242261-92242261 on Build GRCh38
rs749242031 Chr.9: 92242220-92242220 on Build GRCh38
rs562503211 Chr.9: 92242314-92242314 on Build GRCh38
rs769593590 Chr.9: 92242356-92242356 on Build GRCh38
rs775223103 Chr.9: 92242359-92242359 on Build GRCh38
rs767227431 Chr.9: 92242260-92242260 on Build GRCh38
rs370534390 Chr.9: 92242369-92242369 on Build GRCh38
rs771704889 Chr.9: 92242254-92242254 on Build GRCh38
rs760192920 Chr.9: 92242268-92242268 on Build GRCh38
rs1040530991 Chr.9: 92242199-92242199 on Build GRCh38
rs770822119 Chr.9: 92242341-92242341 on Build GRCh38
rs754379616 Chr.9: 92242286-92242286 on Build GRCh38
rs199947184 Chr.9: 92242252-92242252 on Build GRCh38
rs747054053 Chr.9: 92242210-92242210 on Build GRCh38
rs774115743 Chr.9: 92242235-92242235 on Build GRCh38
rs147719645 Chr.9: 92242292-92242292 on Build GRCh38
rs781316563 Chr.9: 92242332-92242332 on Build GRCh38
rs750709544 Chr.9: 92242315-92242315 on Build GRCh38
rs776057292 Chr.9: 92242363-92242363 on Build GRCh38
rs755447093 Chr.9: 92242337-92242338 on Build GRCh38

View Full Product Details