Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 23246765-23247024 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPNMB
Entrez Gene ID
10457
Gene Name
glycoprotein nmb
Gene Aliases
HGFIN, NMB
UniGene
Hs.190495
Gene Chromosome Location
Chr.7: 23246206-23275110 on Build GRCh38
Transcripts
NM_001005340.1 NM_002510.2 XM_005249578.2 XM_017011676.1 XM_017011677.1 XM_017011678.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3832526 c.64G>A p.A22T Chr.7: 23246921-23246921 on Build GRCh38
150248 c.49T>C p.L17L Chr.7: 23246906-23246906 on Build GRCh38
1450053 c.21C>T p.F7F Chr.7: 23246878-23246878 on Build GRCh38
4751208 c.29T>A p.F10Y Chr.7: 23246886-23246886 on Build GRCh38
3880263 c.29T>G p.F10C Chr.7: 23246886-23246886 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs971530338 Chr.7: 23246991-23246991 on Build GRCh38
rs374730555 Chr.7: 23246930-23246930 on Build GRCh38
rs527595866 Chr.7: 23246948-23246948 on Build GRCh38
rs1006840536 Chr.7: 23246928-23246928 on Build GRCh38
rs754508413 Chr.7: 23246845-23246845 on Build GRCh38
rs769787429 Chr.7: 23246807-23246807 on Build GRCh38
rs746783897 Chr.7: 23246801-23246801 on Build GRCh38
rs761042616 Chr.7: 23246842-23246842 on Build GRCh38
rs377587977 Chr.7: 23246962-23246962 on Build GRCh38
rs774966224 Chr.7: 23246828-23246828 on Build GRCh38
rs763770328 Chr.7: 23246840-23246840 on Build GRCh38
rs35519567 Chr.7: 23246906-23246906 on Build GRCh38
rs140122424 Chr.7: 23246910-23246910 on Build GRCh38
rs745718322 Chr.7: 23246895-23246895 on Build GRCh38
rs751202578 Chr.7: 23246841-23246841 on Build GRCh38
rs568434830 Chr.7: 23246999-23246999 on Build GRCh38
rs766855326 Chr.7: 23246844-23246844 on Build GRCh38
rs781443383 Chr.7: 23246966-23246966 on Build GRCh38
rs773987752 Chr.7: 23246922-23246922 on Build GRCh38
rs370923633 Chr.7: 23246967-23246967 on Build GRCh38
rs761368241 Chr.7: 23246927-23246927 on Build GRCh38
rs889962744 Chr.7: 23246959-23246959 on Build GRCh38
rs749289901 Chr.7: 23246804-23246804 on Build GRCh38
rs758442259 Chr.7: 23246870-23246870 on Build GRCh38
rs1025399643 Chr.7: 23246982-23246982 on Build GRCh38
rs150499567 Chr.7: 23246867-23246867 on Build GRCh38
rs748791376 Chr.7: 23246920-23246920 on Build GRCh38
rs760016760 Chr.7: 23246931-23246931 on Build GRCh38
rs758274749 Chr.7: 23246795-23246795 on Build GRCh38
rs771689044 Chr.7: 23246942-23246942 on Build GRCh38
rs111858913 Chr.7: 23246941-23246941 on Build GRCh38
rs115100573 Chr.7: 23246890-23246890 on Build GRCh38
rs201779058 Chr.7: 23246862-23246862 on Build GRCh38
rs979064995 Chr.7: 23246831-23246831 on Build GRCh38
rs140290060 Chr.7: 23246888-23246888 on Build GRCh38
rs937326394 Chr.7: 23246800-23246800 on Build GRCh38
rs992655513 Chr.7: 23246865-23246865 on Build GRCh38
rs779993007 Chr.7: 23246972-23246972 on Build GRCh38
rs17147944 Chr.7: 23246995-23246995 on Build GRCh38
rs369920985 Chr.7: 23246837-23246837 on Build GRCh38
rs778007708 Chr.7: 23246877-23246877 on Build GRCh38
rs370332714 Chr.7: 23246921-23246921 on Build GRCh38
rs980223215 Chr.7: 23246874-23246874 on Build GRCh38
rs747206051 Chr.7: 23246881-23246881 on Build GRCh38
rs192220419 Chr.7: 23246978-23246978 on Build GRCh38
rs201436622 Chr.7: 23246933-23246933 on Build GRCh38
rs1051168932 Chr.7: 23246958-23246958 on Build GRCh38
rs765896670 Chr.7: 23246858-23246858 on Build GRCh38

View Full Product Details