Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 132645811-132646056 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INPP5A
Entrez Gene ID
3632
Gene Name
inositol polyphosphate-5-phosphatase A
Gene Aliases
5PTASE
UniGene
Hs.523360
Gene Chromosome Location
Chr.10: 132537779-132783480 on Build GRCh38
Transcripts
NM_005539.4 XM_017016204.1 XM_017016205.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4012808 c.135G>A p.M45I Chr.10: 132645897-132645897 on Build GRCh38
5743442 c.125C>T p.P42L Chr.10: 132645887-132645887 on Build GRCh38
6128607 c.178A>C p.N60H Chr.10: 132645928-132645928 on Build GRCh38
4880088 c.213C>T p.F71F Chr.10: 132645963-132645963 on Build GRCh38
4144511 c.122A>G p.K41R Chr.10: 132645884-132645884 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777468775 Chr.10: 132645860-132645860 on Build GRCh38
rs199515031 Chr.10: 132645938-132645938 on Build GRCh38
rs375941297 Chr.10: 132645888-132645888 on Build GRCh38
rs374023761 Chr.10: 132645933-132645933 on Build GRCh38
rs771238503 Chr.10: 132645978-132645978 on Build GRCh38
rs570568882 Chr.10: 132645963-132645963 on Build GRCh38
rs200768082 Chr.10: 132645885-132645885 on Build GRCh38
rs144774890 Chr.10: 132645952-132645952 on Build GRCh38
rs112550433 Chr.10: 132645929-132645929 on Build GRCh38
rs748361306 Chr.10: 132645877-132645877 on Build GRCh38
rs752218319 Chr.10: 132645917-132645917 on Build GRCh38
rs371120951 Chr.10: 132645870-132645870 on Build GRCh38
rs547822867 Chr.10: 132645871-132645871 on Build GRCh38
rs777151927 Chr.10: 132645980-132645980 on Build GRCh38
rs768795044 Chr.10: 132645873-132645873 on Build GRCh38
rs748249912 Chr.10: 132645959-132645959 on Build GRCh38
rs767462424 Chr.10: 132646017-132646017 on Build GRCh38
rs376998631 Chr.10: 132645864-132645864 on Build GRCh38
rs147603723 Chr.10: 132646029-132646029 on Build GRCh38
rs568304990 Chr.10: 132645878-132645878 on Build GRCh38
rs747952207 Chr.10: 132645964-132645964 on Build GRCh38
rs774920496 Chr.10: 132645993-132645993 on Build GRCh38
rs58264541 Chr.10: 132645927-132645927 on Build GRCh38
rs370327452 Chr.10: 132645925-132645925 on Build GRCh38
rs752241917 Chr.10: 132645838-132645838 on Build GRCh38
rs925747050 Chr.10: 132645876-132645876 on Build GRCh38
rs762515914 Chr.10: 132646003-132646003 on Build GRCh38
rs761720187 Chr.10: 132646014-132646014 on Build GRCh38
rs750348436 Chr.10: 132645944-132645944 on Build GRCh38
rs917033886 Chr.10: 132645918-132645918 on Build GRCh38
rs562273551 Chr.10: 132645840-132645840 on Build GRCh38
rs140099082 Chr.10: 132645968-132645968 on Build GRCh38
rs1000108920 Chr.10: 132646023-132646023 on Build GRCh38
rs769631811 Chr.10: 132645976-132645976 on Build GRCh38
rs760075203 Chr.10: 132645984-132645984 on Build GRCh38
rs146983957 Chr.10: 132646026-132646026 on Build GRCh38
rs376850593 Chr.10: 132645835-132645835 on Build GRCh38
rs761223770 Chr.10: 132645828-132645828 on Build GRCh38
rs774668350 Chr.10: 132645875-132645875 on Build GRCh38
rs766858771 Chr.10: 132645829-132645829 on Build GRCh38
rs1133400 Chr.10: 132645884-132645884 on Build GRCh38
rs1049348890 Chr.10: 132646026-132646027 on Build GRCh38
rs779488717 Chr.10: 132645834-132645834 on Build GRCh38
rs867758198 Chr.10: 132645894-132645894 on Build GRCh38
rs373265966 Chr.10: 132645879-132645879 on Build GRCh38
rs2274431 Chr.10: 132645989-132645989 on Build GRCh38
rs756050274 Chr.10: 132645951-132645951 on Build GRCh38
rs61757086 Chr.10: 132645924-132645924 on Build GRCh38
rs764610723 Chr.10: 132645895-132645895 on Build GRCh38
rs763655216 Chr.10: 132646012-132646012 on Build GRCh38
rs201359162 Chr.10: 132645858-132645858 on Build GRCh38
rs141407422 Chr.10: 132645837-132645837 on Build GRCh38

View Full Product Details