Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 35875431-35875670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IL7R
Entrez Gene ID
3575
Gene Name
interleukin 7 receptor
Gene Aliases
CD127, CDW127, IL-7R-alpha, IL7RA, ILRA
UniGene
Hs.591742
Gene Chromosome Location
Chr.5: 35856875-35879603 on Build GRCh38
Transcripts
NM_002185.3 NR_120485.1 XM_005248299.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6685457 c.872G>T p.R291I Chr.5: 35875583-35875583 on Build GRCh38
3615751 c.813C>T p.I271I Chr.5: 35875524-35875524 on Build GRCh38
2687261 c.832G>A p.D278N Chr.5: 35875543-35875543 on Build GRCh38
3975511 c.811A>C p.I271L Chr.5: 35875522-35875522 on Build GRCh38
1634208 c.844A>G p.T282A Chr.5: 35875555-35875555 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs766895607 Chr.5: 35875542-35875542 on Build GRCh38
rs752312661 Chr.5: 35875644-35875644 on Build GRCh38
rs866550601 Chr.5: 35875520-35875520 on Build GRCh38
rs747377117 Chr.5: 35875586-35875587 on Build GRCh38
rs775669367 Chr.5: 35875483-35875483 on Build GRCh38
rs113444486 Chr.5: 35875478-35875478 on Build GRCh38
rs986405200 Chr.5: 35875509-35875509 on Build GRCh38
rs199952483 Chr.5: 35875495-35875495 on Build GRCh38
rs921143502 Chr.5: 35875489-35875489 on Build GRCh38
rs200468194 Chr.5: 35875524-35875524 on Build GRCh38
rs199814261 Chr.5: 35875518-35875518 on Build GRCh38
rs768950875 Chr.5: 35875588-35875589 on Build GRCh38
rs987106 Chr.5: 35875491-35875491 on Build GRCh38
rs200058801 Chr.5: 35875618-35875618 on Build GRCh38
rs912324015 Chr.5: 35875512-35875512 on Build GRCh38
rs780909347 Chr.5: 35875475-35875475 on Build GRCh38
rs201449880 Chr.5: 35875543-35875543 on Build GRCh38
rs370063681 Chr.5: 35875624-35875624 on Build GRCh38
rs758984708 Chr.5: 35875617-35875617 on Build GRCh38
rs773614197 Chr.5: 35875522-35875522 on Build GRCh38
rs541039622 Chr.5: 35875612-35875612 on Build GRCh38
rs767908772 Chr.5: 35875499-35875499 on Build GRCh38
rs200896401 Chr.5: 35875630-35875630 on Build GRCh38
rs756911243 Chr.5: 35875595-35875595 on Build GRCh38
rs750270892 Chr.5: 35875612-35875612 on Build GRCh38
rs749310460 Chr.5: 35875640-35875640 on Build GRCh38
rs776380395 Chr.5: 35875629-35875630 on Build GRCh38
rs373309714 Chr.5: 35875556-35875556 on Build GRCh38
rs375571055 Chr.5: 35875639-35875639 on Build GRCh38
rs369971728 Chr.5: 35875525-35875525 on Build GRCh38
rs781012108 Chr.5: 35875601-35875601 on Build GRCh38
rs200927893 Chr.5: 35875477-35875477 on Build GRCh38
rs1036740256 Chr.5: 35875579-35875579 on Build GRCh38
rs1020809522 Chr.5: 35875462-35875462 on Build GRCh38
rs746719598 Chr.5: 35875460-35875460 on Build GRCh38
rs1022245698 Chr.5: 35875564-35875564 on Build GRCh38
rs777036777 Chr.5: 35875595-35875595 on Build GRCh38
rs762007749 Chr.5: 35875498-35875498 on Build GRCh38
rs757074358 Chr.5: 35875464-35875464 on Build GRCh38
rs201897337 Chr.5: 35875633-35875633 on Build GRCh38
rs140034700 Chr.5: 35875487-35875487 on Build GRCh38
rs989564904 Chr.5: 35875545-35875545 on Build GRCh38
rs1024232293 Chr.5: 35875474-35875474 on Build GRCh38
rs898029594 Chr.5: 35875600-35875600 on Build GRCh38
rs375706394 Chr.5: 35875572-35875572 on Build GRCh38

View Full Product Details