Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.2: 226795676-226795929 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IRS1
Entrez Gene ID
3667
Gene Name
insulin receptor substrate 1
Gene Aliases
HIRS-1
UniGene
Hs.471508
Gene Chromosome Location
Chr.2: 226731317-226798790 on Build GRCh38
Transcripts
NM_005544.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5708111 c.2897G>A p.G966D Chr.2: 226795842-226795842 on Build GRCh38
365205 c.2858C>T p.A953V Chr.2: 226795881-226795881 on Build GRCh38
4583266 c.3051A>G p.T1017T Chr.2: 226795688-226795688 on Build GRCh38
6913902 c.3025C>T p.P1009S Chr.2: 226795714-226795714 on Build GRCh38
6457107 c.2827A>C p.K943Q Chr.2: 226795912-226795912 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs979216795 Chr.2: 226795776-226795776 on Build GRCh38
rs776704743 Chr.2: 226795712-226795712 on Build GRCh38
rs777571798 Chr.2: 226795835-226795835 on Build GRCh38
rs772844464 Chr.2: 226795696-226795696 on Build GRCh38
rs972327913 Chr.2: 226795862-226795862 on Build GRCh38
rs967733042 Chr.2: 226795772-226795772 on Build GRCh38
rs765722350 Chr.2: 226795903-226795903 on Build GRCh38
rs745750129 Chr.2: 226795767-226795767 on Build GRCh38
rs751826393 Chr.2: 226795720-226795720 on Build GRCh38
rs766674987 Chr.2: 226795788-226795788 on Build GRCh38
rs780229544 Chr.2: 226795775-226795775 on Build GRCh38
rs761927466 Chr.2: 226795782-226795782 on Build GRCh38
rs746601499 Chr.2: 226795841-226795841 on Build GRCh38
rs757920964 Chr.2: 226795829-226795829 on Build GRCh38
rs1801278 Chr.2: 226795828-226795828 on Build GRCh38
rs143760685 Chr.2: 226795746-226795746 on Build GRCh38
rs371239608 Chr.2: 226795784-226795784 on Build GRCh38
rs369672495 Chr.2: 226795714-226795714 on Build GRCh38
rs373248004 Chr.2: 226795725-226795725 on Build GRCh38
rs758765862 Chr.2: 226795772-226795774 on Build GRCh38
rs141295519 Chr.2: 226795890-226795890 on Build GRCh38
rs748246803 Chr.2: 226795869-226795869 on Build GRCh38
rs750831376 Chr.2: 226795728-226795728 on Build GRCh38
rs771049966 Chr.2: 226795707-226795707 on Build GRCh38
rs1051275735 Chr.2: 226795827-226795827 on Build GRCh38
rs145736083 Chr.2: 226795891-226795891 on Build GRCh38
rs771924555 Chr.2: 226795693-226795693 on Build GRCh38
rs761897696 Chr.2: 226795724-226795724 on Build GRCh38
rs377334234 Chr.2: 226795867-226795867 on Build GRCh38
rs544936782 Chr.2: 226795771-226795771 on Build GRCh38
rs894163217 Chr.2: 226795706-226795706 on Build GRCh38
rs141976450 Chr.2: 226795859-226795859 on Build GRCh38
rs759924294 Chr.2: 226795901-226795901 on Build GRCh38
rs757138873 Chr.2: 226795842-226795842 on Build GRCh38
rs1002889742 Chr.2: 226795899-226795899 on Build GRCh38
rs765744351 Chr.2: 226795809-226795809 on Build GRCh38
rs780609564 Chr.2: 226795735-226795735 on Build GRCh38
rs755401943 Chr.2: 226795799-226795799 on Build GRCh38
rs764021548 Chr.2: 226795717-226795717 on Build GRCh38
rs754945617 Chr.2: 226795790-226795790 on Build GRCh38
rs148431809 Chr.2: 226795801-226795801 on Build GRCh38
rs372034600 Chr.2: 226795739-226795739 on Build GRCh38
rs768810160 Chr.2: 226795768-226795768 on Build GRCh38
rs370373307 Chr.2: 226795896-226795896 on Build GRCh38
rs201935731 Chr.2: 226795802-226795802 on Build GRCh38
rs756462316 Chr.2: 226795730-226795730 on Build GRCh38
rs767686104 Chr.2: 226795783-226795783 on Build GRCh38
rs772126823 Chr.2: 226795888-226795888 on Build GRCh38
rs143731262 Chr.2: 226795895-226795895 on Build GRCh38
rs913587902 Chr.2: 226795753-226795753 on Build GRCh38
rs148150592 Chr.2: 226795766-226795766 on Build GRCh38
rs781090877 Chr.2: 226795858-226795858 on Build GRCh38
rs754367656 Chr.2: 226795793-226795793 on Build GRCh38
rs753180932 Chr.2: 226795904-226795904 on Build GRCh38
rs770949631 Chr.2: 226795740-226795740 on Build GRCh38
rs540624592 Chr.2: 226795736-226795736 on Build GRCh38
rs761036006 Chr.2: 226795787-226795787 on Build GRCh38
rs760515770 Chr.2: 226795703-226795703 on Build GRCh38
rs373195857 Chr.2: 226795837-226795837 on Build GRCh38
rs988057825 Chr.2: 226795750-226795750 on Build GRCh38
rs377465384 Chr.2: 226795758-226795758 on Build GRCh38
rs747866903 Chr.2: 226795742-226795742 on Build GRCh38
rs367762669 Chr.2: 226795747-226795747 on Build GRCh38

View Full Product Details