Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.21: 31945969-31946240 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HUNK
Entrez Gene ID
30811
Gene Name
hormonally up-regulated Neu-associated kinase
Gene Aliases
-
UniGene
Hs.109437
Gene Chromosome Location
Chr.21: 31873315-32004064 on Build GRCh38
Transcripts
NM_014586.1 XM_011529537.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6329896 c.688G>T p.A230S Chr.21: 31946113-31946113 on Build GRCh38
4101228 c.706G>A p.A236T Chr.21: 31946131-31946131 on Build GRCh38
4506893 c.732C>T p.I244I Chr.21: 31946157-31946157 on Build GRCh38
6004709 c.690T>C p.A230A Chr.21: 31946115-31946115 on Build GRCh38
5567725 c.662G>A p.S221N Chr.21: 31946087-31946087 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs374957204 Chr.21: 31946131-31946131 on Build GRCh38
rs374009656 Chr.21: 31946194-31946194 on Build GRCh38
rs925106010 Chr.21: 31946200-31946200 on Build GRCh38
rs1014549869 Chr.21: 31946056-31946056 on Build GRCh38
rs777598067 Chr.21: 31946005-31946005 on Build GRCh38
rs1005916664 Chr.21: 31946028-31946028 on Build GRCh38
rs992502321 Chr.21: 31946081-31946081 on Build GRCh38
rs765737663 Chr.21: 31946069-31946069 on Build GRCh38
rs768342149 Chr.21: 31946133-31946133 on Build GRCh38
rs771376041 Chr.21: 31946152-31946152 on Build GRCh38
rs752968893 Chr.21: 31946102-31946102 on Build GRCh38
rs757152359 Chr.21: 31946109-31946109 on Build GRCh38
rs770894265 Chr.21: 31946009-31946009 on Build GRCh38
rs767753828 Chr.21: 31946086-31946086 on Build GRCh38
rs771853330 Chr.21: 31946024-31946024 on Build GRCh38
rs751597883 Chr.21: 31946080-31946080 on Build GRCh38
rs766851869 Chr.21: 31946077-31946077 on Build GRCh38
rs759574531 Chr.21: 31946082-31946082 on Build GRCh38
rs140621578 Chr.21: 31946169-31946169 on Build GRCh38
rs368005070 Chr.21: 31946048-31946048 on Build GRCh38
rs375377891 Chr.21: 31946006-31946006 on Build GRCh38
rs774875015 Chr.21: 31946157-31946157 on Build GRCh38
rs763429549 Chr.21: 31946076-31946076 on Build GRCh38
rs760942096 Chr.21: 31946199-31946199 on Build GRCh38
rs773788877 Chr.21: 31946019-31946019 on Build GRCh38
rs764316051 Chr.21: 31946202-31946202 on Build GRCh38
rs763312357 Chr.21: 31946139-31946139 on Build GRCh38
rs371506151 Chr.21: 31946118-31946118 on Build GRCh38
rs371936194 Chr.21: 31946000-31946000 on Build GRCh38
rs75293794 Chr.21: 31946103-31946103 on Build GRCh38
rs762290307 Chr.21: 31946061-31946061 on Build GRCh38
rs745323358 Chr.21: 31946020-31946020 on Build GRCh38
rs200605258 Chr.21: 31946216-31946216 on Build GRCh38
rs777491361 Chr.21: 31946107-31946107 on Build GRCh38
rs773464178 Chr.21: 31946135-31946135 on Build GRCh38
rs932949742 Chr.21: 31945994-31945994 on Build GRCh38
rs766332712 Chr.21: 31946112-31946112 on Build GRCh38
rs779688008 Chr.21: 31946128-31946128 on Build GRCh38
rs773621924 Chr.21: 31946075-31946075 on Build GRCh38
rs367617331 Chr.21: 31946184-31946184 on Build GRCh38
rs747970408 Chr.21: 31945991-31945991 on Build GRCh38
rs961649041 Chr.21: 31946058-31946058 on Build GRCh38
rs980652882 Chr.21: 31946188-31946188 on Build GRCh38
rs16988660 Chr.21: 31946127-31946127 on Build GRCh38
rs760018148 Chr.21: 31946180-31946180 on Build GRCh38

View Full Product Details