Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 15119885-15120158 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ILVBL
Entrez Gene ID
10994
Gene Name
ilvB acetolactate synthase like
Gene Aliases
209L8, AHAS, HACL1L, ILV2H
UniGene
Hs.78880
Gene Chromosome Location
Chr.19: 15114974-15125799 on Build GRCh38
Transcripts
NM_006844.4 XM_005259717.4 XM_011527651.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4642263 c.809C>T p.P270L Chr.19: 15119989-15119989 on Build GRCh38
5484240 c.779C>T p.P260L Chr.19: 15120019-15120019 on Build GRCh38
4645327 c.783G>A p.L261L Chr.19: 15120015-15120015 on Build GRCh38
4567577 c.770_771CC>TT p.P257L Chr.19: 15120027-15120028 on Build GRCh38
5657758 c.760delG p.E254fs*27 Chr.19: 15120038-15120038 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs970676008 Chr.19: 15119912-15119912 on Build GRCh38
rs1029438796 Chr.19: 15120128-15120128 on Build GRCh38
rs943119084 Chr.19: 15120010-15120010 on Build GRCh38
rs547285334 Chr.19: 15120065-15120065 on Build GRCh38
rs922421750 Chr.19: 15119933-15119933 on Build GRCh38
rs1038773117 Chr.19: 15120011-15120011 on Build GRCh38
rs997919317 Chr.19: 15120126-15120126 on Build GRCh38
rs953886969 Chr.19: 15119965-15119965 on Build GRCh38
rs201923875 Chr.19: 15120023-15120023 on Build GRCh38
rs956183260 Chr.19: 15119992-15119992 on Build GRCh38
rs985760689 Chr.19: 15119976-15119976 on Build GRCh38
rs935927822 Chr.19: 15119995-15119995 on Build GRCh38
rs773015464 Chr.19: 15119920-15119920 on Build GRCh38
rs773156514 Chr.19: 15120003-15120003 on Build GRCh38
rs886686424 Chr.19: 15120059-15120059 on Build GRCh38
rs867949653 Chr.19: 15119946-15119946 on Build GRCh38
rs115852958 Chr.19: 15120019-15120019 on Build GRCh38
rs200136775 Chr.19: 15119998-15119998 on Build GRCh38
rs559852629 Chr.19: 15120026-15120026 on Build GRCh38
rs750926212 Chr.19: 15120095-15120095 on Build GRCh38
rs888670330 Chr.19: 15120086-15120086 on Build GRCh38
rs201670039 Chr.19: 15119989-15119989 on Build GRCh38
rs895333075 Chr.19: 15120116-15120116 on Build GRCh38
rs559684946 Chr.19: 15120038-15120038 on Build GRCh38
rs1015256726 Chr.19: 15120100-15120100 on Build GRCh38
rs1037854927 Chr.19: 15120113-15120113 on Build GRCh38
rs775641218 Chr.19: 15119996-15119996 on Build GRCh38
rs978083424 Chr.19: 15119954-15119954 on Build GRCh38
rs1012385050 Chr.19: 15120127-15120127 on Build GRCh38
rs543919477 Chr.19: 15119937-15119937 on Build GRCh38
rs759340634 Chr.19: 15119949-15119949 on Build GRCh38
rs374867337 Chr.19: 15120092-15120092 on Build GRCh38
rs913932896 Chr.19: 15120018-15120018 on Build GRCh38
rs770885943 Chr.19: 15119932-15119932 on Build GRCh38
rs181709901 Chr.19: 15120027-15120027 on Build GRCh38

View Full Product Details