Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 89418439-89418696 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
IFIT5
Entrez Gene ID
24138
Gene Name
interferon induced protein with tetratricopeptide repeats 5
Gene Aliases
ISG58, P58, RI58
UniGene
Hs.252839
Gene Chromosome Location
Chr.10: 89414568-89421002 on Build GRCh38
Transcripts
NM_012420.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3807940 c.1434C>G p.L478L Chr.10: 89418633-89418633 on Build GRCh38
4451518 c.1264A>C p.K422Q Chr.10: 89418463-89418463 on Build GRCh38
6683847 c.1272T>C p.S424S Chr.10: 89418471-89418471 on Build GRCh38
6258301 c.1303G>T p.V435L Chr.10: 89418502-89418502 on Build GRCh38
4466614 c.1443C>T p.S481S Chr.10: 89418642-89418642 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751942622 Chr.10: 89418613-89418613 on Build GRCh38
rs375747724 Chr.10: 89418619-89418619 on Build GRCh38
rs763808953 Chr.10: 89418466-89418466 on Build GRCh38
rs752936937 Chr.10: 89418619-89418629 on Build GRCh38
rs745670236 Chr.10: 89418654-89418654 on Build GRCh38
rs756223280 Chr.10: 89418491-89418491 on Build GRCh38
rs749460798 Chr.10: 89418532-89418532 on Build GRCh38
rs745661525 Chr.10: 89418561-89418561 on Build GRCh38
rs756634022 Chr.10: 89418635-89418635 on Build GRCh38
rs761581329 Chr.10: 89418472-89418472 on Build GRCh38
rs757410092 Chr.10: 89418651-89418651 on Build GRCh38
rs1035985112 Chr.10: 89418620-89418620 on Build GRCh38
rs777085281 Chr.10: 89418464-89418464 on Build GRCh38
rs370049282 Chr.10: 89418621-89418621 on Build GRCh38
rs759888870 Chr.10: 89418622-89418622 on Build GRCh38
rs373375883 Chr.10: 89418479-89418479 on Build GRCh38
rs781611318 Chr.10: 89418483-89418483 on Build GRCh38
rs761491449 Chr.10: 89418599-89418599 on Build GRCh38
rs760241604 Chr.10: 89418573-89418573 on Build GRCh38
rs768041597 Chr.10: 89418624-89418624 on Build GRCh38
rs755458159 Chr.10: 89418480-89418480 on Build GRCh38
rs776313935 Chr.10: 89418589-89418589 on Build GRCh38
rs202194886 Chr.10: 89418521-89418521 on Build GRCh38
rs142594899 Chr.10: 89418602-89418602 on Build GRCh38
rs777787123 Chr.10: 89418643-89418643 on Build GRCh38
rs763387850 Chr.10: 89418634-89418635 on Build GRCh38
rs753594215 Chr.10: 89418468-89418468 on Build GRCh38
rs758177764 Chr.10: 89418656-89418656 on Build GRCh38
rs772032781 Chr.10: 89418563-89418563 on Build GRCh38
rs753188686 Chr.10: 89418626-89418626 on Build GRCh38
rs761969166 Chr.10: 89418485-89418485 on Build GRCh38
rs753917343 Chr.10: 89418650-89418650 on Build GRCh38
rs568835542 Chr.10: 89418565-89418565 on Build GRCh38
rs367854137 Chr.10: 89418658-89418658 on Build GRCh38
rs1043389423 Chr.10: 89418517-89418517 on Build GRCh38
rs896294106 Chr.10: 89418657-89418657 on Build GRCh38
rs771171026 Chr.10: 89418540-89418540 on Build GRCh38
rs770093124 Chr.10: 89418554-89418554 on Build GRCh38
rs189548496 Chr.10: 89418652-89418652 on Build GRCh38
rs537774876 Chr.10: 89418584-89418584 on Build GRCh38
rs765623484 Chr.10: 89418587-89418590 on Build GRCh38

View Full Product Details