Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.20: 62218566-62218791 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HRH3
Entrez Gene ID
11255
Gene Name
histamine receptor H3
Gene Aliases
GPCR97, HH3R
UniGene
Hs.251399
Gene Chromosome Location
Chr.20: 62213845-62220267 on Build GRCh38
Transcripts
NM_007232.2 XM_005260266.3 XM_017027623.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
232504 c.271T>C p.Y91H Chr.20: 62218637-62218637 on Build GRCh38
6417944 c.259T>G p.C87G Chr.20: 62218649-62218649 on Build GRCh38
6300721 c.314G>T p.G105V Chr.20: 62218594-62218594 on Build GRCh38
3693613 c.253G>A p.A85T Chr.20: 62218655-62218655 on Build GRCh38
6417943 c.259T>G p.C87G Chr.20: 62218649-62218649 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs201299080 Chr.20: 62218675-62218675 on Build GRCh38
rs779001130 Chr.20: 62218613-62218613 on Build GRCh38
rs577219078 Chr.20: 62218731-62218731 on Build GRCh38
rs201073853 Chr.20: 62218706-62218706 on Build GRCh38
rs202004300 Chr.20: 62218597-62218597 on Build GRCh38
rs745912908 Chr.20: 62218623-62218623 on Build GRCh38
rs200469744 Chr.20: 62218669-62218669 on Build GRCh38
rs1052194475 Chr.20: 62218722-62218722 on Build GRCh38
rs200313117 Chr.20: 62218621-62218621 on Build GRCh38
rs775684112 Chr.20: 62218641-62218641 on Build GRCh38
rs867597398 Chr.20: 62218644-62218644 on Build GRCh38
rs765689427 Chr.20: 62218693-62218693 on Build GRCh38
rs776984289 Chr.20: 62218662-62218662 on Build GRCh38
rs201585083 Chr.20: 62218601-62218601 on Build GRCh38
rs773822203 Chr.20: 62218698-62218698 on Build GRCh38
rs201117959 Chr.20: 62218602-62218602 on Build GRCh38
rs199591850 Chr.20: 62218680-62218680 on Build GRCh38
rs765928291 Chr.20: 62218593-62218593 on Build GRCh38
rs769818909 Chr.20: 62218733-62218733 on Build GRCh38
rs199633132 Chr.20: 62218656-62218656 on Build GRCh38
rs201390271 Chr.20: 62218635-62218635 on Build GRCh38
rs754804647 Chr.20: 62218598-62218598 on Build GRCh38
rs915027069 Chr.20: 62218723-62218723 on Build GRCh38
rs201373345 Chr.20: 62218595-62218595 on Build GRCh38
rs747290921 Chr.20: 62218655-62218655 on Build GRCh38
rs762342098 Chr.20: 62218670-62218670 on Build GRCh38
rs970352387 Chr.20: 62218745-62218745 on Build GRCh38
rs768202907 Chr.20: 62218643-62218643 on Build GRCh38
rs915255498 Chr.20: 62218712-62218712 on Build GRCh38
rs999251752 Chr.20: 62218758-62218758 on Build GRCh38
rs200538029 Chr.20: 62218687-62218687 on Build GRCh38
rs35075005 Chr.20: 62218716-62218716 on Build GRCh38
rs947533082 Chr.20: 62218734-62218734 on Build GRCh38
rs764705251 Chr.20: 62218691-62218697 on Build GRCh38
rs202146230 Chr.20: 62218612-62218612 on Build GRCh38

View Full Product Details