Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 122848599-122848872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HIP1R
Entrez Gene ID
9026
Gene Name
huntingtin interacting protein 1 related
Gene Aliases
HIP12, HIP3, ILWEQ
UniGene
Hs.524815
Gene Chromosome Location
Chr.12: 122834498-122862961 on Build GRCh38
Transcripts
NM_001303097.1 NM_001303099.1 NM_003959.2 XM_011538963.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3687858 c.351C>T p.D117D Chr.12: 122848846-122848846 on Build GRCh38
7139751 c.338G>T p.R113L Chr.12: 122848833-122848833 on Build GRCh38
4544477 c.357G>A p.W119* Chr.12: 122848852-122848852 on Build GRCh38
6282825 c.337C>T p.R113W Chr.12: 122848832-122848832 on Build GRCh38
4039679 c.326G>A p.R109H Chr.12: 122848821-122848821 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs747978801 Chr.12: 122848656-122848656 on Build GRCh38
rs757074821 Chr.12: 122848723-122848723 on Build GRCh38
rs763448852 Chr.12: 122848838-122848838 on Build GRCh38
rs368574693 Chr.12: 122848762-122848762 on Build GRCh38
rs776773873 Chr.12: 122848753-122848753 on Build GRCh38
rs555407898 Chr.12: 122848659-122848659 on Build GRCh38
rs764726502 Chr.12: 122848768-122848768 on Build GRCh38
rs771064907 Chr.12: 122848820-122848820 on Build GRCh38
rs776776573 Chr.12: 122848827-122848827 on Build GRCh38
rs777995898 Chr.12: 122848665-122848665 on Build GRCh38
rs758671133 Chr.12: 122848637-122848637 on Build GRCh38
rs759940826 Chr.12: 122848754-122848754 on Build GRCh38
rs770074345 Chr.12: 122848832-122848832 on Build GRCh38
rs182805687 Chr.12: 122848833-122848833 on Build GRCh38
rs747076283 Chr.12: 122848815-122848815 on Build GRCh38
rs754676538 Chr.12: 122848778-122848778 on Build GRCh38
rs1003611186 Chr.12: 122848733-122848733 on Build GRCh38
rs879628027 Chr.12: 122848841-122848841 on Build GRCh38
rs755784941 Chr.12: 122848638-122848638 on Build GRCh38
rs530896606 Chr.12: 122848814-122848814 on Build GRCh38
rs367731331 Chr.12: 122848626-122848626 on Build GRCh38
rs191071864 Chr.12: 122848667-122848667 on Build GRCh38
rs571167825 Chr.12: 122848732-122848732 on Build GRCh38
rs758729996 Chr.12: 122848705-122848705 on Build GRCh38
rs757076268 Chr.12: 122848668-122848668 on Build GRCh38
rs1021454823 Chr.12: 122848783-122848783 on Build GRCh38
rs771968327 Chr.12: 122848790-122848790 on Build GRCh38
rs1010543975 Chr.12: 122848731-122848731 on Build GRCh38
rs952810050 Chr.12: 122848849-122848849 on Build GRCh38
rs752182295 Chr.12: 122848847-122848847 on Build GRCh38
rs868615533 Chr.12: 122848852-122848852 on Build GRCh38
rs758921640 Chr.12: 122848764-122848764 on Build GRCh38
rs972875935 Chr.12: 122848799-122848799 on Build GRCh38
rs774203549 Chr.12: 122848654-122848654 on Build GRCh38
rs551658487 Chr.12: 122848728-122848728 on Build GRCh38
rs921415772 Chr.12: 122848691-122848691 on Build GRCh38
rs749098815 Chr.12: 122848646-122848646 on Build GRCh38
rs372034414 Chr.12: 122848846-122848846 on Build GRCh38
rs749919060 Chr.12: 122848771-122848771 on Build GRCh38
rs533111320 Chr.12: 122848714-122848714 on Build GRCh38
rs753210833 Chr.12: 122848619-122848619 on Build GRCh38
rs924286336 Chr.12: 122848831-122848831 on Build GRCh38
rs74487507 Chr.12: 122848699-122848699 on Build GRCh38
rs779755733 Chr.12: 122848772-122848772 on Build GRCh38
rs768406136 Chr.12: 122848651-122848651 on Build GRCh38
rs186167211 Chr.12: 122848658-122848658 on Build GRCh38
rs759614978 Chr.12: 122848830-122848830 on Build GRCh38
rs746738271 Chr.12: 122848667-122848667 on Build GRCh38
rs201535038 Chr.12: 122848760-122848760 on Build GRCh38
rs764687556 Chr.12: 122848840-122848840 on Build GRCh38
rs901001185 Chr.12: 122848666-122848666 on Build GRCh38
rs1040843718 Chr.12: 122848670-122848670 on Build GRCh38
rs748006593 Chr.12: 122848789-122848789 on Build GRCh38
rs2292133 Chr.12: 122848657-122848657 on Build GRCh38
rs557803190 Chr.12: 122848632-122848632 on Build GRCh38
rs368178583 Chr.12: 122848620-122848621 on Build GRCh38
rs376804189 Chr.12: 122848700-122848700 on Build GRCh38
rs778524497 Chr.12: 122848788-122848788 on Build GRCh38

View Full Product Details