Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.19: 18436631-18436872 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ISYNA1
Entrez Gene ID
51477
Gene Name
inositol-3-phosphate synthase 1
Gene Aliases
INO1, INOS, IPS, IPS 1, IPS-1
UniGene
Hs.405873
Gene Chromosome Location
Chr.19: 18434388-18438301 on Build GRCh38
Transcripts
NM_001170938.1 NM_001253389.1 NM_016368.4 NR_045573.1 NR_045574.1 XM_011528059.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5162563 c.576G>A p.A192A Chr.19: 18436717-18436717 on Build GRCh38
6437347 c.555G>A p.A185A Chr.19: 18436738-18436738 on Build GRCh38
4693196 c.450G>A p.A150A Chr.19: 18436843-18436843 on Build GRCh38
6043252 c.492A>C p.Q164H Chr.19: 18436801-18436801 on Build GRCh38
1391685 c.501G>A p.P167P Chr.19: 18436792-18436792 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs778890208 Chr.19: 18436717-18436717 on Build GRCh38
rs774117364 Chr.19: 18436852-18436852 on Build GRCh38
rs368377568 Chr.19: 18436657-18436657 on Build GRCh38
rs767903366 Chr.19: 18436759-18436759 on Build GRCh38
rs756524230 Chr.19: 18436773-18436773 on Build GRCh38
rs745849041 Chr.19: 18436837-18436837 on Build GRCh38
rs1029172674 Chr.19: 18436673-18436673 on Build GRCh38
rs993231647 Chr.19: 18436812-18436812 on Build GRCh38
rs762410485 Chr.19: 18436730-18436733 on Build GRCh38
rs374113839 Chr.19: 18436691-18436691 on Build GRCh38
rs780972489 Chr.19: 18436831-18436831 on Build GRCh38
rs898375630 Chr.19: 18436792-18436792 on Build GRCh38
rs775096976 Chr.19: 18436726-18436726 on Build GRCh38
rs748877705 Chr.19: 18436848-18436848 on Build GRCh38
rs147482450 Chr.19: 18436690-18436690 on Build GRCh38
rs752408483 Chr.19: 18436653-18436653 on Build GRCh38
rs917606458 Chr.19: 18436750-18436750 on Build GRCh38
rs762430471 Chr.19: 18436654-18436654 on Build GRCh38
rs764857591 Chr.19: 18436717-18436718 on Build GRCh38
rs763638072 Chr.19: 18436655-18436655 on Build GRCh38
rs773743984 Chr.19: 18436741-18436741 on Build GRCh38
rs755265547 Chr.19: 18436785-18436785 on Build GRCh38
rs762629715 Chr.19: 18436735-18436735 on Build GRCh38
rs762311401 Chr.19: 18436754-18436754 on Build GRCh38
rs755077720 Chr.19: 18436713-18436713 on Build GRCh38
rs770779084 Chr.19: 18436719-18436719 on Build GRCh38
rs749293912 Chr.19: 18436762-18436762 on Build GRCh38
rs1055154489 Chr.19: 18436670-18436670 on Build GRCh38
rs775289903 Chr.19: 18436843-18436843 on Build GRCh38
rs540468469 Chr.19: 18436665-18436665 on Build GRCh38
rs766889926 Chr.19: 18436776-18436776 on Build GRCh38
rs745650840 Chr.19: 18436723-18436723 on Build GRCh38
rs144717477 Chr.19: 18436676-18436676 on Build GRCh38
rs776699315 Chr.19: 18436720-18436720 on Build GRCh38
rs759242237 Chr.19: 18436690-18436716 on Build GRCh38
rs199555469 Chr.19: 18436815-18436815 on Build GRCh38
rs974986104 Chr.19: 18436663-18436663 on Build GRCh38
rs752248602 Chr.19: 18436716-18436717 on Build GRCh38
rs529148937 Chr.19: 18436718-18436718 on Build GRCh38
rs769628350 Chr.19: 18436724-18436724 on Build GRCh38
rs376639419 Chr.19: 18436849-18436849 on Build GRCh38
rs200584853 Chr.19: 18436796-18436796 on Build GRCh38
rs754073061 Chr.19: 18436781-18436781 on Build GRCh38
rs148517966 Chr.19: 18436804-18436804 on Build GRCh38
rs367647209 Chr.19: 18436697-18436697 on Build GRCh38

View Full Product Details