Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 68313830-68314055 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITGA11
Entrez Gene ID
22801
Gene Name
integrin subunit alpha 11
Gene Aliases
HsT18964
UniGene
Hs.436416
Gene Chromosome Location
Chr.15: 68297433-68432312 on Build GRCh38
Transcripts
NM_001004439.1 XM_005254228.3 XM_011521363.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4693227 c.2806C>T p.R936W Chr.15: 68313855-68313855 on Build GRCh38
1239156 c.2829C>T p.N943N Chr.15: 68313832-68313832 on Build GRCh38
2257884 c.2803G>T p.E935* Chr.15: 68313858-68313858 on Build GRCh38
2257883 c.2804A>T p.E935V Chr.15: 68313857-68313857 on Build GRCh38
1285593 c.2793-2A>T p.? Chr.15: 68313870-68313870 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs970844516 Chr.15: 68314034-68314034 on Build GRCh38
rs750877284 Chr.15: 68313892-68313892 on Build GRCh38
rs996890660 Chr.15: 68313946-68313946 on Build GRCh38
rs529371107 Chr.15: 68314011-68314011 on Build GRCh38
rs760235690 Chr.15: 68313887-68313887 on Build GRCh38
rs909986410 Chr.15: 68313974-68313974 on Build GRCh38
rs971298161 Chr.15: 68313923-68313923 on Build GRCh38
rs988478912 Chr.15: 68313906-68313906 on Build GRCh38
rs765915060 Chr.15: 68313890-68313890 on Build GRCh38
rs368307734 Chr.15: 68313854-68313854 on Build GRCh38
rs930521165 Chr.15: 68313959-68313959 on Build GRCh38
rs758800892 Chr.15: 68313902-68313902 on Build GRCh38
rs952850031 Chr.15: 68313996-68313996 on Build GRCh38
rs766583123 Chr.15: 68313911-68313911 on Build GRCh38
rs901215191 Chr.15: 68314000-68314000 on Build GRCh38
rs201167323 Chr.15: 68313886-68313886 on Build GRCh38
rs984581522 Chr.15: 68313963-68313963 on Build GRCh38
rs111957377 Chr.15: 68313933-68313933 on Build GRCh38
rs751836638 Chr.15: 68313916-68313916 on Build GRCh38
rs781207165 Chr.15: 68313853-68313853 on Build GRCh38
rs769771137 Chr.15: 68313855-68313855 on Build GRCh38
rs543958930 Chr.15: 68313989-68313989 on Build GRCh38
rs868079589 Chr.15: 68314018-68314018 on Build GRCh38
rs767161650 Chr.15: 68313881-68313881 on Build GRCh38
rs983257835 Chr.15: 68314005-68314005 on Build GRCh38
rs1050040089 Chr.15: 68314024-68314024 on Build GRCh38
rs927099352 Chr.15: 68313953-68313953 on Build GRCh38
rs755086418 Chr.15: 68313850-68313850 on Build GRCh38
rs572655465 Chr.15: 68313856-68313856 on Build GRCh38
rs371353194 Chr.15: 68313859-68313859 on Build GRCh38
rs558106186 Chr.15: 68313924-68313924 on Build GRCh38
rs370711647 Chr.15: 68313992-68313992 on Build GRCh38
rs769660864 Chr.15: 68313890-68313891 on Build GRCh38
rs570517606 Chr.15: 68313863-68313863 on Build GRCh38
rs774060103 Chr.15: 68313864-68313864 on Build GRCh38
rs1015775095 Chr.15: 68314022-68314022 on Build GRCh38

View Full Product Details