Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.7: 27155942-27156203 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
HOXA7
Entrez Gene ID
3204
Gene Name
homeobox A7
Gene Aliases
ANTP, HOX1, HOX1.1, HOX1A
UniGene
Hs.610216
Gene Chromosome Location
Chr.7: 27153716-27156677 on Build GRCh38
Transcripts
NM_006896.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3027270 c.364T>A p.W122R Chr.7: 27156182-27156182 on Build GRCh38
6413231 c.370C>T p.R124W Chr.7: 27156176-27156176 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs558046500 Chr.7: 27156138-27156138 on Build GRCh38
rs902685416 Chr.7: 27155983-27155983 on Build GRCh38
rs570565406 Chr.7: 27156022-27156022 on Build GRCh38
rs757142588 Chr.7: 27156145-27156145 on Build GRCh38
rs753006970 Chr.7: 27156129-27156129 on Build GRCh38
rs1033804182 Chr.7: 27156050-27156050 on Build GRCh38
rs1014684805 Chr.7: 27156183-27156183 on Build GRCh38
rs1049397206 Chr.7: 27156116-27156116 on Build GRCh38
rs775283215 Chr.7: 27156044-27156044 on Build GRCh38
rs930473785 Chr.7: 27156109-27156110 on Build GRCh38
rs996182724 Chr.7: 27156094-27156108 on Build GRCh38
rs994025385 Chr.7: 27156100-27156100 on Build GRCh38
rs544001319 Chr.7: 27156175-27156175 on Build GRCh38
rs751357018 Chr.7: 27156141-27156141 on Build GRCh38
rs572595353 Chr.7: 27156052-27156052 on Build GRCh38
rs539512953 Chr.7: 27156024-27156024 on Build GRCh38
rs1012347748 Chr.7: 27156027-27156027 on Build GRCh38
rs188491382 Chr.7: 27156105-27156105 on Build GRCh38
rs1056458407 Chr.7: 27155991-27155991 on Build GRCh38
rs887788215 Chr.7: 27156040-27156040 on Build GRCh38
rs1033251050 Chr.7: 27156076-27156076 on Build GRCh38
rs779567620 Chr.7: 27156184-27156184 on Build GRCh38
rs886352106 Chr.7: 27156124-27156124 on Build GRCh38
rs763815863 Chr.7: 27156139-27156139 on Build GRCh38
rs960780142 Chr.7: 27156096-27156096 on Build GRCh38
rs895788001 Chr.7: 27156154-27156154 on Build GRCh38
rs956309040 Chr.7: 27156077-27156077 on Build GRCh38
rs1023479152 Chr.7: 27156073-27156074 on Build GRCh38
rs755689640 Chr.7: 27156182-27156182 on Build GRCh38
rs896552911 Chr.7: 27156020-27156020 on Build GRCh38
rs375834510 Chr.7: 27156148-27156148 on Build GRCh38
rs865788066 Chr.7: 27156176-27156176 on Build GRCh38
rs961186416 Chr.7: 27156074-27156074 on Build GRCh38
rs553021877 Chr.7: 27156036-27156036 on Build GRCh38
rs921694125 Chr.7: 27156106-27156106 on Build GRCh38
rs555244138 Chr.7: 27156126-27156126 on Build GRCh38
rs900297061 Chr.7: 27155987-27155987 on Build GRCh38
rs988472505 Chr.7: 27156085-27156091 on Build GRCh38
rs935532999 Chr.7: 27155988-27155988 on Build GRCh38
rs979358073 Chr.7: 27156075-27156088 on Build GRCh38
rs765358859 Chr.7: 27156122-27156122 on Build GRCh38
rs944359497 Chr.7: 27156094-27156094 on Build GRCh38

View Full Product Details