Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 24336497-24336752 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRHL3
Entrez Gene ID
57822
Gene Name
grainyhead like transcription factor 3
Gene Aliases
SOM, TFCP2L4, VWS2
UniGene
Hs.657920
Gene Chromosome Location
Chr.1: 24319322-24364482 on Build GRCh38
Transcripts
NM_001195010.1 NM_021180.3 NM_198173.2 NM_198174.2 XM_011541869.1 XM_011541870.2

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4804795 c.386C>A p.P129Q Chr.1: 24336586-24336586 on Build GRCh38
6051373 c.403delT p.L135fs*78 Chr.1: 24336618-24336618 on Build GRCh38
2052002 c.415T>G p.L139V Chr.1: 24336630-24336630 on Build GRCh38
4751356 c.546C>G p.N182K Chr.1: 24336746-24336746 on Build GRCh38
2051997 c.317C>T p.T106I Chr.1: 24336517-24336517 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs150037319 Chr.1: 24336566-24336566 on Build GRCh38
rs763050512 Chr.1: 24336620-24336620 on Build GRCh38
rs770927831 Chr.1: 24336554-24336554 on Build GRCh38
rs773199434 Chr.1: 24336695-24336695 on Build GRCh38
rs140960821 Chr.1: 24336660-24336660 on Build GRCh38
rs147738765 Chr.1: 24336628-24336628 on Build GRCh38
rs186914295 Chr.1: 24336564-24336564 on Build GRCh38
rs767040060 Chr.1: 24336629-24336629 on Build GRCh38
rs778111145 Chr.1: 24336654-24336654 on Build GRCh38
rs779025609 Chr.1: 24336671-24336671 on Build GRCh38
rs34637004 Chr.1: 24336694-24336694 on Build GRCh38
rs749812508 Chr.1: 24336536-24336536 on Build GRCh38
rs774380464 Chr.1: 24336724-24336724 on Build GRCh38
rs773225729 Chr.1: 24336586-24336586 on Build GRCh38
rs774357267 Chr.1: 24336555-24336555 on Build GRCh38
rs144050104 Chr.1: 24336728-24336728 on Build GRCh38
rs765145591 Chr.1: 24336571-24336571 on Build GRCh38
rs994104716 Chr.1: 24336640-24336640 on Build GRCh38
rs939598160 Chr.1: 24336627-24336627 on Build GRCh38
rs908953686 Chr.1: 24336602-24336602 on Build GRCh38
rs148080626 Chr.1: 24336642-24336642 on Build GRCh38
rs369259121 Chr.1: 24336633-24336633 on Build GRCh38
rs752284034 Chr.1: 24336731-24336731 on Build GRCh38
rs762962991 Chr.1: 24336714-24336714 on Build GRCh38
rs376281609 Chr.1: 24336621-24336621 on Build GRCh38
rs751080248 Chr.1: 24336625-24336625 on Build GRCh38
rs758924862 Chr.1: 24336725-24336725 on Build GRCh38
rs111363131 Chr.1: 24336563-24336563 on Build GRCh38
rs146706821 Chr.1: 24336550-24336550 on Build GRCh38
rs371889064 Chr.1: 24336687-24336687 on Build GRCh38
rs139093980 Chr.1: 24336686-24336686 on Build GRCh38
rs756415680 Chr.1: 24336648-24336648 on Build GRCh38
rs756505220 Chr.1: 24336529-24336529 on Build GRCh38
rs35868286 Chr.1: 24336722-24336722 on Build GRCh38
rs929115407 Chr.1: 24336539-24336539 on Build GRCh38
rs757817057 Chr.1: 24336668-24336668 on Build GRCh38
rs761658802 Chr.1: 24336567-24336567 on Build GRCh38
rs778208867 Chr.1: 24336532-24336532 on Build GRCh38

View Full Product Details