Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 94838402-94838675 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
INTS8
Entrez Gene ID
55656
Gene Name
integrator complex subunit 8
Gene Aliases
C8orf52, INT8
UniGene
Hs.727669
Gene Chromosome Location
Chr.8: 94823290-94894664 on Build GRCh38
Transcripts
NM_017864.3 NR_073444.1 NR_073445.1 XM_017013616.1 XM_017013617.1 XM_017013618.1 XM_017013619.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
5938213 c.887C>T p.T296I Chr.8: 94838488-94838488 on Build GRCh38
3652016 c.974C>T p.P325L Chr.8: 94838575-94838575 on Build GRCh38
5776847 c.880C>T p.H294Y Chr.8: 94838481-94838481 on Build GRCh38
7325792 c.901C>T p.R301W Chr.8: 94838502-94838502 on Build GRCh38
3367440 c.991A>G p.I331V Chr.8: 94838592-94838592 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs755022101 Chr.8: 94838560-94838560 on Build GRCh38
rs773670124 Chr.8: 94838646-94838646 on Build GRCh38
rs550816890 Chr.8: 94838596-94838596 on Build GRCh38
rs770533677 Chr.8: 94838495-94838495 on Build GRCh38
rs570684449 Chr.8: 94838611-94838611 on Build GRCh38
rs781125023 Chr.8: 94838467-94838467 on Build GRCh38
rs77629237 Chr.8: 94838474-94838474 on Build GRCh38
rs1038924226 Chr.8: 94838552-94838552 on Build GRCh38
rs752623504 Chr.8: 94838564-94838564 on Build GRCh38
rs770573037 Chr.8: 94838590-94838590 on Build GRCh38
rs780157797 Chr.8: 94838441-94838441 on Build GRCh38
rs751609337 Chr.8: 94838557-94838557 on Build GRCh38
rs1029711387 Chr.8: 94838609-94838609 on Build GRCh38
rs773875408 Chr.8: 94838502-94838502 on Build GRCh38
rs777515344 Chr.8: 94838580-94838580 on Build GRCh38
rs758995001 Chr.8: 94838504-94838504 on Build GRCh38
rs777283578 Chr.8: 94838525-94838525 on Build GRCh38
rs945056345 Chr.8: 94838473-94838473 on Build GRCh38
rs754903556 Chr.8: 94838455-94838455 on Build GRCh38
rs748920409 Chr.8: 94838491-94838491 on Build GRCh38
rs763202526 Chr.8: 94838539-94838539 on Build GRCh38
rs750865401 Chr.8: 94838534-94838534 on Build GRCh38
rs762386295 Chr.8: 94838629-94838629 on Build GRCh38
rs533143946 Chr.8: 94838621-94838621 on Build GRCh38
rs530790185 Chr.8: 94838540-94838540 on Build GRCh38
rs749072336 Chr.8: 94838584-94838584 on Build GRCh38
rs562023912 Chr.8: 94838517-94838517 on Build GRCh38
rs975799330 Chr.8: 94838605-94838605 on Build GRCh38
rs368747539 Chr.8: 94838446-94838446 on Build GRCh38
rs756094745 Chr.8: 94838574-94838574 on Build GRCh38
rs770463171 Chr.8: 94838633-94838633 on Build GRCh38
rs903795214 Chr.8: 94838516-94838516 on Build GRCh38
rs765686329 Chr.8: 94838531-94838531 on Build GRCh38
rs965791781 Chr.8: 94838604-94838604 on Build GRCh38
rs762478454 Chr.8: 94838530-94838530 on Build GRCh38
rs774022803 Chr.8: 94838642-94838642 on Build GRCh38
rs1007270215 Chr.8: 94838545-94838545 on Build GRCh38
rs771705955 Chr.8: 94838614-94838614 on Build GRCh38
rs769579185 Chr.8: 94838476-94838476 on Build GRCh38
rs777609276 Chr.8: 94838489-94838489 on Build GRCh38

View Full Product Details