Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.3: 141807584-141807817 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GRK7
Entrez Gene ID
131890
Gene Name
G protein-coupled receptor kinase 7
Gene Aliases
GPRK7
UniGene
Hs.680654
Gene Chromosome Location
Chr.3: 141763127-141817373 on Build GRCh38
Transcripts
NM_139209.2 XM_017005717.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6675816 c.1057A>G p.T353A Chr.3: 141807651-141807651 on Build GRCh38
5355072 c.1197T>C p.S399S Chr.3: 141807791-141807791 on Build GRCh38
3588220 c.1163G>A p.R388Q Chr.3: 141807757-141807757 on Build GRCh38
5355190 c.1220C>T p.T407I Chr.3: 141807814-141807814 on Build GRCh38
5578676 c.1133G>A p.G378E Chr.3: 141807727-141807727 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs879070744 Chr.3: 141807663-141807663 on Build GRCh38
rs781674398 Chr.3: 141807623-141807623 on Build GRCh38
rs751748396 Chr.3: 141807716-141807716 on Build GRCh38
rs372832933 Chr.3: 141807644-141807644 on Build GRCh38
rs201809353 Chr.3: 141807693-141807693 on Build GRCh38
rs765459838 Chr.3: 141807690-141807690 on Build GRCh38
rs151124237 Chr.3: 141807746-141807746 on Build GRCh38
rs540433932 Chr.3: 141807633-141807633 on Build GRCh38
rs776739277 Chr.3: 141807656-141807656 on Build GRCh38
rs775724780 Chr.3: 141807781-141807781 on Build GRCh38
rs892955316 Chr.3: 141807613-141807613 on Build GRCh38
rs746616025 Chr.3: 141807761-141807761 on Build GRCh38
rs541213789 Chr.3: 141807701-141807701 on Build GRCh38
rs141435292 Chr.3: 141807786-141807786 on Build GRCh38
rs143817637 Chr.3: 141807704-141807704 on Build GRCh38
rs768609112 Chr.3: 141807767-141807767 on Build GRCh38
rs199515525 Chr.3: 141807757-141807757 on Build GRCh38
rs753244680 Chr.3: 141807734-141807734 on Build GRCh38
rs755272627 Chr.3: 141807619-141807619 on Build GRCh38
rs964856000 Chr.3: 141807733-141807733 on Build GRCh38
rs890372504 Chr.3: 141807724-141807724 on Build GRCh38
rs778549653 Chr.3: 141807739-141807739 on Build GRCh38
rs748607779 Chr.3: 141807629-141807629 on Build GRCh38
rs772370456 Chr.3: 141807622-141807622 on Build GRCh38
rs751868148 Chr.3: 141807617-141807617 on Build GRCh38
rs776541822 Chr.3: 141807773-141807773 on Build GRCh38
rs745592263 Chr.3: 141807635-141807635 on Build GRCh38
rs766517806 Chr.3: 141807705-141807705 on Build GRCh38
rs111477566 Chr.3: 141807632-141807632 on Build GRCh38
rs758530186 Chr.3: 141807610-141807610 on Build GRCh38
rs746791533 Chr.3: 141807646-141807646 on Build GRCh38
rs766829180 Chr.3: 141807614-141807614 on Build GRCh38
rs768519134 Chr.3: 141807648-141807648 on Build GRCh38
rs748157258 Chr.3: 141807776-141807776 on Build GRCh38
rs199678773 Chr.3: 141807645-141807645 on Build GRCh38
rs756599305 Chr.3: 141807737-141807737 on Build GRCh38
rs767830659 Chr.3: 141807726-141807726 on Build GRCh38
rs778364461 Chr.3: 141807634-141807634 on Build GRCh38
rs141035095 Chr.3: 141807756-141807756 on Build GRCh38

View Full Product Details