Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 90625057-90625330 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADGRV1
Entrez Gene ID
84059
Gene Name
adhesion G protein-coupled receptor V1
Gene Aliases
FEB4, GPR98, MASS1, USH2B, USH2C, VLGR1, VLGR1b
UniGene
Hs.591777
Gene Chromosome Location
Chr.5: 90558681-91164226 on Build GRCh38
Transcripts
NM_032119.3 NR_003149.1 XM_017009963.1 XM_017009964.1 XM_017009965.1 XM_017009966.1 XM_017009967.1 XM_017009968.1 XM_017009969.1 XM_017009970.1 XM_017009971.1 XM_017009974.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3357916 c.582T>C p.P194P Chr.5: 90625153-90625153 on Build GRCh38
738966 c.669C>T p.D223D Chr.5: 90625240-90625240 on Build GRCh38
4586070 c.560T>G p.V187G Chr.5: 90625131-90625131 on Build GRCh38
5415710 c.672+1G>T p.? Chr.5: 90625244-90625244 on Build GRCh38
6104604 c.562G>T p.E188* Chr.5: 90625133-90625133 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs751425835 Chr.5: 90625096-90625096 on Build GRCh38
rs757180527 Chr.5: 90625116-90625116 on Build GRCh38
rs758132314 Chr.5: 90625087-90625087 on Build GRCh38
rs760159666 Chr.5: 90625185-90625185 on Build GRCh38
rs61745496 Chr.5: 90625167-90625167 on Build GRCh38
rs369418418 Chr.5: 90625140-90625140 on Build GRCh38
rs756977835 Chr.5: 90625193-90625193 on Build GRCh38
rs775156018 Chr.5: 90625193-90625193 on Build GRCh38
rs555037066 Chr.5: 90625273-90625273 on Build GRCh38
rs758576790 Chr.5: 90625269-90625269 on Build GRCh38
rs192654926 Chr.5: 90625145-90625145 on Build GRCh38
rs767405355 Chr.5: 90625249-90625249 on Build GRCh38
rs748303111 Chr.5: 90625148-90625148 on Build GRCh38
rs776978539 Chr.5: 90625171-90625171 on Build GRCh38
rs762649515 Chr.5: 90625230-90625230 on Build GRCh38
rs935013327 Chr.5: 90625278-90625278 on Build GRCh38
rs772293155 Chr.5: 90625149-90625149 on Build GRCh38
rs369189825 Chr.5: 90625084-90625084 on Build GRCh38
rs749017926 Chr.5: 90625133-90625136 on Build GRCh38
rs746306894 Chr.5: 90625284-90625284 on Build GRCh38
rs747403211 Chr.5: 90625163-90625163 on Build GRCh38
rs763927162 Chr.5: 90625091-90625091 on Build GRCh38
rs781626159 Chr.5: 90625281-90625281 on Build GRCh38
rs374946426 Chr.5: 90625240-90625240 on Build GRCh38
rs876657474 Chr.5: 90625211-90625211 on Build GRCh38
rs916797724 Chr.5: 90625231-90625231 on Build GRCh38
rs756980621 Chr.5: 90625245-90625245 on Build GRCh38
rs373287553 Chr.5: 90625111-90625111 on Build GRCh38
rs930241382 Chr.5: 90625196-90625196 on Build GRCh38
rs750322737 Chr.5: 90625250-90625250 on Build GRCh38
rs950784228 Chr.5: 90625131-90625131 on Build GRCh38
rs746071555 Chr.5: 90625179-90625179 on Build GRCh38
rs200207490 Chr.5: 90625294-90625294 on Build GRCh38
rs780962333 Chr.5: 90625130-90625130 on Build GRCh38
rs764590195 Chr.5: 90625191-90625191 on Build GRCh38
rs377529304 Chr.5: 90625135-90625135 on Build GRCh38
rs989503177 Chr.5: 90625115-90625115 on Build GRCh38
rs957459495 Chr.5: 90625110-90625110 on Build GRCh38
rs41311331 Chr.5: 90625299-90625299 on Build GRCh38
rs727504958 Chr.5: 90625241-90625241 on Build GRCh38
rs1032835066 Chr.5: 90625097-90625097 on Build GRCh38
rs748248200 Chr.5: 90625259-90625259 on Build GRCh38
rs778904250 Chr.5: 90625255-90625255 on Build GRCh38
rs61745498 Chr.5: 90625152-90625152 on Build GRCh38
rs910039292 Chr.5: 90625271-90625271 on Build GRCh38
rs755504936 Chr.5: 90625085-90625085 on Build GRCh38
rs535773920 Chr.5: 90625270-90625270 on Build GRCh38
rs763872010 Chr.5: 90625235-90625235 on Build GRCh38

View Full Product Details