Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 63183464-63183715 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JMJD1C
Entrez Gene ID
221037
Gene Name
jumonji domain containing 1C
Gene Aliases
TRIP-8, TRIP8
UniGene
Hs.413416
Gene Chromosome Location
Chr.10: 63167221-63522075 on Build GRCh38
Transcripts
NM_001282948.1 NM_001318153.1 NM_001318154.1 NM_001322252.1 NM_001322254.1 NM_001322258.1 NM_032776.2 NR_134512.1 XM_011539503.1 XM_017015897.1 XM_017015898.1 XM_017015899.1 XM_017015900.1 XM_017015901.1 XM_017015902.1 XM_017015903.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2158356 c.6962-1G>A p.? Chr.10: 63183570-63183570 on Build GRCh38
5733006 c.6338G>A p.G2113D Chr.10: 63183482-63183482 on Build GRCh38
5033627 c.6320A>G p.N2107S Chr.10: 63183500-63183500 on Build GRCh38
6689563 c.7033A>G p.I2345V Chr.10: 63183498-63183498 on Build GRCh38
6689555 c.6315_6317delTGT p.V2106delV Chr.10: 63183503-63183505 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs780428642 Chr.10: 63183493-63183493 on Build GRCh38
rs543620801 Chr.10: 63183600-63183600 on Build GRCh38
rs748348204 Chr.10: 63183534-63183534 on Build GRCh38
rs761544238 Chr.10: 63183617-63183617 on Build GRCh38
rs948975924 Chr.10: 63183648-63183648 on Build GRCh38
rs772578294 Chr.10: 63183677-63183677 on Build GRCh38
rs957306224 Chr.10: 63183682-63183682 on Build GRCh38
rs910022164 Chr.10: 63183660-63183660 on Build GRCh38
rs763947371 Chr.10: 63183607-63183607 on Build GRCh38
rs561080043 Chr.10: 63183651-63183651 on Build GRCh38
rs865890342 Chr.10: 63183598-63183598 on Build GRCh38
rs749335937 Chr.10: 63183553-63183553 on Build GRCh38
rs917294127 Chr.10: 63183641-63183641 on Build GRCh38
rs770063370 Chr.10: 63183541-63183541 on Build GRCh38
rs375596153 Chr.10: 63183618-63183618 on Build GRCh38
rs771989931 Chr.10: 63183575-63183575 on Build GRCh38
rs746161035 Chr.10: 63183573-63183573 on Build GRCh38
rs151183153 Chr.10: 63183636-63183636 on Build GRCh38
rs781577975 Chr.10: 63183533-63183533 on Build GRCh38
rs747351059 Chr.10: 63183520-63183520 on Build GRCh38
rs749880729 Chr.10: 63183619-63183619 on Build GRCh38
rs531366793 Chr.10: 63183543-63183543 on Build GRCh38
rs773278691 Chr.10: 63183549-63183549 on Build GRCh38
rs779633334 Chr.10: 63183621-63183621 on Build GRCh38
rs775309287 Chr.10: 63183577-63183577 on Build GRCh38
rs372703563 Chr.10: 63183601-63183601 on Build GRCh38
rs897961335 Chr.10: 63183609-63183609 on Build GRCh38
rs776613135 Chr.10: 63183608-63183608 on Build GRCh38
rs755330672 Chr.10: 63183526-63183526 on Build GRCh38

View Full Product Details