Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 23569409-23569670 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
JPH4
Entrez Gene ID
84502
Gene Name
junctophilin 4
Gene Aliases
JP4, JPHL1
UniGene
Hs.597841
Gene Chromosome Location
Chr.14: 23568035-23578800 on Build GRCh38
Transcripts
NM_001146028.1 NM_032452.2

SNP Detail

SNP ID Chromosome Location
rs570454345 Chr.14: 23569499-23569499 on Build GRCh38
rs142956727 Chr.14: 23569499-23569500 on Build GRCh38
rs201491387 Chr.14: 23569500-23569500 on Build GRCh38
rs939876888 Chr.14: 23569603-23569603 on Build GRCh38
rs912014005 Chr.14: 23569492-23569492 on Build GRCh38
rs1048060282 Chr.14: 23569574-23569574 on Build GRCh38
rs1001303250 Chr.14: 23569496-23569499 on Build GRCh38
rs377476544 Chr.14: 23569600-23569600 on Build GRCh38
rs995383205 Chr.14: 23569524-23569527 on Build GRCh38
rs375140240 Chr.14: 23569483-23569483 on Build GRCh38
rs932294895 Chr.14: 23569507-23569507 on Build GRCh38
rs373149920 Chr.14: 23569599-23569599 on Build GRCh38
rs1045404178 Chr.14: 23569487-23569488 on Build GRCh38
rs745861907 Chr.14: 23569588-23569588 on Build GRCh38
rs571578919 Chr.14: 23569492-23569495 on Build GRCh38
rs760356106 Chr.14: 23569650-23569650 on Build GRCh38
rs905637465 Chr.14: 23569488-23569488 on Build GRCh38
rs71413993 Chr.14: 23569504-23569504 on Build GRCh38
rs547157990 Chr.14: 23569484-23569484 on Build GRCh38
rs556831950 Chr.14: 23569501-23569501 on Build GRCh38
rs989026056 Chr.14: 23569487-23569487 on Build GRCh38
rs865975704 Chr.14: 23569489-23569489 on Build GRCh38
rs772863535 Chr.14: 23569540-23569540 on Build GRCh38
rs533927273 Chr.14: 23569452-23569452 on Build GRCh38

View Full Product Details