Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.4: 185162727-185162998 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CFAP97
Entrez Gene ID
57587
Gene Name
cilia and flagella associated protein 97
Gene Aliases
KIAA1430, hmw
UniGene
Hs.535734
Gene Chromosome Location
Chr.4: 185159662-185210251 on Build GRCh38
Transcripts
NM_020827.2 XM_017008483.1 XM_017008484.1 XM_017008485.1 XM_017008486.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4468403 c.1541C>T p.S514F Chr.4: 185162856-185162856 on Build GRCh38
1594495 c.1529C>T p.A510V Chr.4: 185162868-185162868 on Build GRCh38
6435855 c.1597T>C p.*533Q Chr.4: 185162800-185162800 on Build GRCh38
4409687 c.1472-4T>G p.? Chr.4: 185162929-185162929 on Build GRCh38
4645511 c.1555C>T p.R519* Chr.4: 185162842-185162842 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs779939237 Chr.4: 185162842-185162842 on Build GRCh38
rs767760582 Chr.4: 185162962-185162964 on Build GRCh38
rs748333073 Chr.4: 185162777-185162777 on Build GRCh38
rs977921581 Chr.4: 185162869-185162869 on Build GRCh38
rs746967089 Chr.4: 185162948-185162948 on Build GRCh38
rs924007539 Chr.4: 185162890-185162890 on Build GRCh38
rs372936990 Chr.4: 185162835-185162835 on Build GRCh38
rs764696170 Chr.4: 185162833-185162833 on Build GRCh38
rs369524420 Chr.4: 185162815-185162815 on Build GRCh38
rs772863623 Chr.4: 185162935-185162935 on Build GRCh38
rs376547220 Chr.4: 185162814-185162814 on Build GRCh38
rs370139008 Chr.4: 185162933-185162933 on Build GRCh38
rs374928103 Chr.4: 185162862-185162862 on Build GRCh38
rs192418727 Chr.4: 185162868-185162868 on Build GRCh38
rs542600963 Chr.4: 185162780-185162780 on Build GRCh38
rs761406692 Chr.4: 185162921-185162921 on Build GRCh38
rs920776494 Chr.4: 185162966-185162966 on Build GRCh38
rs772384761 Chr.4: 185162809-185162809 on Build GRCh38
rs770973387 Chr.4: 185162800-185162800 on Build GRCh38
rs960383619 Chr.4: 185162871-185162871 on Build GRCh38
rs760075022 Chr.4: 185162808-185162808 on Build GRCh38
rs770988835 Chr.4: 185162956-185162956 on Build GRCh38
rs776320379 Chr.4: 185162915-185162915 on Build GRCh38
rs372535407 Chr.4: 185162900-185162900 on Build GRCh38
rs754797898 Chr.4: 185162857-185162857 on Build GRCh38
rs775761727 Chr.4: 185162886-185162886 on Build GRCh38
rs778042401 Chr.4: 185162858-185162858 on Build GRCh38
rs761367687 Chr.4: 185162826-185162826 on Build GRCh38
rs573901906 Chr.4: 185162764-185162764 on Build GRCh38
rs766415704 Chr.4: 185162972-185162972 on Build GRCh38
rs964960135 Chr.4: 185162794-185162794 on Build GRCh38
rs774817164 Chr.4: 185162801-185162801 on Build GRCh38
rs746322555 Chr.4: 185162872-185162872 on Build GRCh38
rs775852032 Chr.4: 185162812-185162812 on Build GRCh38
rs368858532 Chr.4: 185162754-185162754 on Build GRCh38
rs895156591 Chr.4: 185162950-185162950 on Build GRCh38
rs772578555 Chr.4: 185162874-185162874 on Build GRCh38
rs758433480 Chr.4: 185162841-185162841 on Build GRCh38
rs531111986 Chr.4: 185162941-185162941 on Build GRCh38
rs1022088388 Chr.4: 185162849-185162849 on Build GRCh38
rs751405435 Chr.4: 185162847-185162847 on Build GRCh38
rs146037638 Chr.4: 185162867-185162867 on Build GRCh38
rs939759121 Chr.4: 185162922-185162922 on Build GRCh38
rs61729128 Chr.4: 185162901-185162901 on Build GRCh38
rs777648405 Chr.4: 185162784-185162784 on Build GRCh38
rs1050652578 Chr.4: 185162970-185162970 on Build GRCh38
rs753495178 Chr.4: 185162823-185162823 on Build GRCh38
rs781482826 Chr.4: 185162969-185162969 on Build GRCh38
rs76868819 Chr.4: 185162792-185162792 on Build GRCh38

View Full Product Details