Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 74328316-74328567 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
KIF19
Entrez Gene ID
124602
Gene Name
kinesin family member 19
Gene Aliases
KIF19A
UniGene
Hs.372773
Gene Chromosome Location
Chr.17: 74326212-74355820 on Build GRCh38
Transcripts
NM_153209.3 XM_005257029.1 XM_005257030.1 XM_017024150.1 XM_017024151.1 XM_017024152.1 XM_017024153.1 XM_017024154.1 XM_017024155.1 XM_017024156.1 XM_017024157.1 XM_017024158.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
1385740 c.66C>T p.S22S Chr.17: 74328451-74328451 on Build GRCh38
984014 c.120+5_120+6insC p.? Chr.17: 74328510-74328511 on Build GRCh38
5665810 c.77T>A p.L26Q Chr.17: 74328462-74328462 on Build GRCh38
3890325 c.103C>T p.H35Y Chr.17: 74328488-74328488 on Build GRCh38
4917305 c.80A>G p.E27G Chr.17: 74328465-74328465 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs756106187 Chr.17: 74328477-74328477 on Build GRCh38
rs777911654 Chr.17: 74328484-74328484 on Build GRCh38
rs746362776 Chr.17: 74328430-74328430 on Build GRCh38
rs898195063 Chr.17: 74328482-74328482 on Build GRCh38
rs555062106 Chr.17: 74328351-74328351 on Build GRCh38
rs747747135 Chr.17: 74328539-74328539 on Build GRCh38
rs190406158 Chr.17: 74328460-74328460 on Build GRCh38
rs772685901 Chr.17: 74328508-74328508 on Build GRCh38
rs75263687 Chr.17: 74328449-74328449 on Build GRCh38
rs147059711 Chr.17: 74328437-74328437 on Build GRCh38
rs758348730 Chr.17: 74328387-74328387 on Build GRCh38
rs752811158 Chr.17: 74328399-74328399 on Build GRCh38
rs988838274 Chr.17: 74328400-74328400 on Build GRCh38
rs757556936 Chr.17: 74328500-74328500 on Build GRCh38
rs768238018 Chr.17: 74328440-74328440 on Build GRCh38
rs776222447 Chr.17: 74328532-74328532 on Build GRCh38
rs747553968 Chr.17: 74328520-74328520 on Build GRCh38
rs28673468 Chr.17: 74328402-74328402 on Build GRCh38
rs1020566516 Chr.17: 74328491-74328491 on Build GRCh38
rs1009865725 Chr.17: 74328488-74328488 on Build GRCh38
rs77996437 Chr.17: 74328446-74328446 on Build GRCh38
rs780659563 Chr.17: 74328518-74328518 on Build GRCh38
rs79421135 Chr.17: 74328447-74328447 on Build GRCh38
rs953569757 Chr.17: 74328370-74328370 on Build GRCh38
rs553967354 Chr.17: 74328452-74328452 on Build GRCh38
rs746274829 Chr.17: 74328507-74328507 on Build GRCh38
rs1035946499 Chr.17: 74328368-74328368 on Build GRCh38
rs761535250 Chr.17: 74328441-74328441 on Build GRCh38
rs372238416 Chr.17: 74328512-74328512 on Build GRCh38
rs74753097 Chr.17: 74328540-74328540 on Build GRCh38
rs143033806 Chr.17: 74328394-74328394 on Build GRCh38
rs750298696 Chr.17: 74328381-74328381 on Build GRCh38
rs751516664 Chr.17: 74328473-74328473 on Build GRCh38
rs370336937 Chr.17: 74328429-74328429 on Build GRCh38
rs905634846 Chr.17: 74328439-74328439 on Build GRCh38
rs543148965 Chr.17: 74328489-74328489 on Build GRCh38
rs377445897 Chr.17: 74328521-74328521 on Build GRCh38
rs11412158 Chr.17: 74328510-74328511 on Build GRCh38
rs147705520 Chr.17: 74328451-74328451 on Build GRCh38
rs374844997 Chr.17: 74328513-74328513 on Build GRCh38
rs753949157 Chr.17: 74328485-74328485 on Build GRCh38
rs762684759 Chr.17: 74328547-74328547 on Build GRCh38
rs749687448 Chr.17: 74328460-74328460 on Build GRCh38
rs397800366 Chr.17: 74328511-74328512 on Build GRCh38
rs370793314 Chr.17: 74328435-74328435 on Build GRCh38
rs779415708 Chr.17: 74328506-74328506 on Build GRCh38
rs987528262 Chr.17: 74328375-74328375 on Build GRCh38
rs140463696 Chr.17: 74328434-74328434 on Build GRCh38
rs749555101 Chr.17: 74328416-74328416 on Build GRCh38
rs765114277 Chr.17: 74328376-74328376 on Build GRCh38
rs568873108 Chr.17: 74328364-74328364 on Build GRCh38
rs772965578 Chr.17: 74328541-74328541 on Build GRCh38

View Full Product Details