Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 48258726-48258999 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ITM2B
Entrez Gene ID
9445
Gene Name
integral membrane protein 2B
Gene Aliases
ABRI, BRI, BRI2, BRICD2B, E25B, E3-16, FBD, RDGCA, imBRI2
UniGene
Hs.643683
Gene Chromosome Location
Chr.13: 48233138-48262096 on Build GRCh38
Transcripts
NM_021999.4

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
357508 c.664C>T p.L222L Chr.13: 48258896-48258896 on Build GRCh38
1300222 c.583C>G p.Q195E Chr.13: 48258815-48258815 on Build GRCh38
1367114 c.605A>G p.H202R Chr.13: 48258837-48258837 on Build GRCh38
947778 c.618_635del18 p.R208_D213delRIENID Chr.13: 48258850-48258867 on Build GRCh38
947779 c.623G>A p.R208H Chr.13: 48258855-48258855 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs373587210 Chr.13: 48258817-48258817 on Build GRCh38
rs199884227 Chr.13: 48258894-48258894 on Build GRCh38
rs748421647 Chr.13: 48258772-48258772 on Build GRCh38
rs11556901 Chr.13: 48258815-48258815 on Build GRCh38
rs763629732 Chr.13: 48258920-48258920 on Build GRCh38
rs201852577 Chr.13: 48258820-48258820 on Build GRCh38
rs773922518 Chr.13: 48258869-48258869 on Build GRCh38
rs750219288 Chr.13: 48258763-48258763 on Build GRCh38
rs760214237 Chr.13: 48258914-48258914 on Build GRCh38
rs757237881 Chr.13: 48258756-48258756 on Build GRCh38
rs767295680 Chr.13: 48258759-48258759 on Build GRCh38
rs767045938 Chr.13: 48258893-48258893 on Build GRCh38
rs775381567 Chr.13: 48258777-48258777 on Build GRCh38
rs368050477 Chr.13: 48258930-48258930 on Build GRCh38
rs756025247 Chr.13: 48258929-48258929 on Build GRCh38
rs1048587576 Chr.13: 48258883-48258883 on Build GRCh38
rs761159398 Chr.13: 48258891-48258891 on Build GRCh38
rs201399334 Chr.13: 48258855-48258855 on Build GRCh38
rs150094093 Chr.13: 48258766-48258766 on Build GRCh38
rs777940570 Chr.13: 48258790-48258790 on Build GRCh38
rs777802039 Chr.13: 48258946-48258946 on Build GRCh38
rs889966863 Chr.13: 48258923-48258923 on Build GRCh38
rs368554353 Chr.13: 48258754-48258754 on Build GRCh38
rs778253495 Chr.13: 48258797-48258810 on Build GRCh38
rs370211396 Chr.13: 48258765-48258765 on Build GRCh38
rs762725137 Chr.13: 48258854-48258854 on Build GRCh38
rs1007037150 Chr.13: 48258928-48258928 on Build GRCh38
rs770332292 Chr.13: 48258822-48258822 on Build GRCh38
rs918883589 Chr.13: 48258814-48258814 on Build GRCh38
rs775963867 Chr.13: 48258838-48258838 on Build GRCh38
rs753591191 Chr.13: 48258937-48258937 on Build GRCh38
rs903269829 Chr.13: 48258857-48258857 on Build GRCh38
rs371745063 Chr.13: 48258942-48258942 on Build GRCh38
rs747013966 Chr.13: 48258810-48258810 on Build GRCh38
rs747117971 Chr.13: 48258971-48258971 on Build GRCh38
rs1042075786 Chr.13: 48258856-48258856 on Build GRCh38

View Full Product Details