Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.5: 151025311-151025546 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPX3
Entrez Gene ID
2878
Gene Name
glutathione peroxidase 3
Gene Aliases
GPx-P, GSHPx-3, GSHPx-P
UniGene
Hs.386793
Gene Chromosome Location
Chr.5: 151020438-151028993 on Build GRCh38
Transcripts
NM_002084.3

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6674724 c.129C>T p.Y43Y Chr.5: 151025381-151025381 on Build GRCh38
3244553 c.190G>A p.V64I Chr.5: 151025442-151025442 on Build GRCh38
6169965 c.205G>A p.V69M Chr.5: 151025457-151025457 on Build GRCh38
6298620 c.103G>T p.G35C Chr.5: 151025355-151025355 on Build GRCh38
3919381 c.124G>A p.E42K Chr.5: 151025376-151025376 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs79763168 Chr.5: 151025337-151025337 on Build GRCh38
rs757369344 Chr.5: 151025500-151025502 on Build GRCh38
rs901489884 Chr.5: 151025395-151025395 on Build GRCh38
rs770916140 Chr.5: 151025483-151025483 on Build GRCh38
rs775560970 Chr.5: 151025489-151025489 on Build GRCh38
rs772010208 Chr.5: 151025382-151025382 on Build GRCh38
rs773392965 Chr.5: 151025379-151025379 on Build GRCh38
rs751560977 Chr.5: 151025436-151025437 on Build GRCh38
rs142849792 Chr.5: 151025456-151025456 on Build GRCh38
rs371856135 Chr.5: 151025479-151025479 on Build GRCh38
rs997240109 Chr.5: 151025411-151025411 on Build GRCh38
rs748297956 Chr.5: 151025469-151025469 on Build GRCh38
rs753136454 Chr.5: 151025404-151025404 on Build GRCh38
rs766813820 Chr.5: 151025441-151025441 on Build GRCh38
rs760476705 Chr.5: 151025397-151025397 on Build GRCh38
rs367725712 Chr.5: 151025351-151025351 on Build GRCh38
rs748438053 Chr.5: 151025375-151025375 on Build GRCh38
rs761543657 Chr.5: 151025518-151025518 on Build GRCh38
rs764034885 Chr.5: 151025413-151025413 on Build GRCh38
rs749907550 Chr.5: 151025431-151025431 on Build GRCh38
rs764043839 Chr.5: 151025400-151025400 on Build GRCh38
rs764620936 Chr.5: 151025415-151025415 on Build GRCh38
rs374183847 Chr.5: 151025333-151025333 on Build GRCh38
rs769022356 Chr.5: 151025354-151025354 on Build GRCh38
rs200128541 Chr.5: 151025376-151025376 on Build GRCh38
rs757867437 Chr.5: 151025433-151025433 on Build GRCh38
rs371966819 Chr.5: 151025405-151025405 on Build GRCh38
rs200228989 Chr.5: 151025480-151025480 on Build GRCh38
rs373981887 Chr.5: 151025455-151025455 on Build GRCh38
rs994657894 Chr.5: 151025513-151025513 on Build GRCh38
rs911673697 Chr.5: 151025352-151025352 on Build GRCh38
rs368521899 Chr.5: 151025491-151025491 on Build GRCh38
rs776606918 Chr.5: 151025516-151025516 on Build GRCh38
rs781596538 Chr.5: 151025457-151025457 on Build GRCh38
rs781326577 Chr.5: 151025366-151025366 on Build GRCh38
rs768547423 Chr.5: 151025509-151025509 on Build GRCh38
rs756402136 Chr.5: 151025478-151025478 on Build GRCh38
rs780569082 Chr.5: 151025348-151025348 on Build GRCh38
rs890034936 Chr.5: 151025341-151025341 on Build GRCh38
rs759363512 Chr.5: 151025381-151025381 on Build GRCh38
rs1027597893 Chr.5: 151025519-151025519 on Build GRCh38
rs959323331 Chr.5: 151025511-151025511 on Build GRCh38
rs775577576 Chr.5: 151025396-151025396 on Build GRCh38
rs762659510 Chr.5: 151025402-151025404 on Build GRCh38

View Full Product Details