Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 66924123-66924362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GPHN
Entrez Gene ID
10243
Gene Name
gephyrin
Gene Aliases
GEPH, GPH, GPHRYN, HKPX1, MOCODC
UniGene
Hs.208765
Gene Chromosome Location
Chr.14: 66507407-67735831 on Build GRCh38
Transcripts
NM_001024218.1 NM_020806.4 XM_005267254.3 XM_011536340.2 XM_011536342.2 XM_011536343.2 XM_011536344.2 XM_011536345.2 XM_011536346.2 XM_011536347.2 XM_017020913.1 XM_017020914.1 XM_017020915.1 XM_017020916.1 XM_017020917.1 XM_017020918.1 XM_017020919.1 XM_017020920.1 XM_017020921.1 XM_017020922.1 XM_017020923.1 XM_017020924.1 XM_017020925.1 XM_017020926.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
287189 c.817A>G p.T273A Chr.14: 66924281-66924281 on Build GRCh38
957125 c.826C>T p.R276W Chr.14: 66924290-66924290 on Build GRCh38
6673257 c.827G>T p.R276L Chr.14: 66924291-66924291 on Build GRCh38
5924617 c.787C>T p.P263S Chr.14: 66924251-66924251 on Build GRCh38
281444 c.780G>A p.Q260Q Chr.14: 66924244-66924244 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs960957890 Chr.14: 66924278-66924278 on Build GRCh38
rs543454860 Chr.14: 66924185-66924185 on Build GRCh38
rs771609374 Chr.14: 66924282-66924282 on Build GRCh38
rs753042559 Chr.14: 66924161-66924162 on Build GRCh38
rs773971749 Chr.14: 66924272-66924272 on Build GRCh38
rs575826167 Chr.14: 66924208-66924208 on Build GRCh38
rs765574246 Chr.14: 66924175-66924175 on Build GRCh38
rs1001175732 Chr.14: 66924180-66924180 on Build GRCh38
rs966844291 Chr.14: 66924316-66924316 on Build GRCh38
rs756515605 Chr.14: 66924167-66924168 on Build GRCh38
rs199511542 Chr.14: 66924260-66924260 on Build GRCh38
rs767565175 Chr.14: 66924339-66924339 on Build GRCh38
rs766600695 Chr.14: 66924195-66924195 on Build GRCh38
rs751726828 Chr.14: 66924215-66924215 on Build GRCh38
rs561779418 Chr.14: 66924235-66924235 on Build GRCh38
rs777009433 Chr.14: 66924290-66924290 on Build GRCh38
rs41285470 Chr.14: 66924264-66924264 on Build GRCh38
rs745440766 Chr.14: 66924275-66924275 on Build GRCh38
rs200818040 Chr.14: 66924332-66924332 on Build GRCh38
rs988027321 Chr.14: 66924299-66924299 on Build GRCh38
rs748108654 Chr.14: 66924231-66924231 on Build GRCh38
rs755083907 Chr.14: 66924227-66924227 on Build GRCh38
rs964081476 Chr.14: 66924228-66924228 on Build GRCh38
rs369062570 Chr.14: 66924163-66924163 on Build GRCh38
rs533003321 Chr.14: 66924293-66924293 on Build GRCh38
rs781045927 Chr.14: 66924230-66924230 on Build GRCh38
rs762152734 Chr.14: 66924172-66924172 on Build GRCh38
rs764346460 Chr.14: 66924300-66924301 on Build GRCh38
rs201387623 Chr.14: 66924313-66924313 on Build GRCh38
rs763382762 Chr.14: 66924318-66924318 on Build GRCh38
rs765544216 Chr.14: 66924301-66924301 on Build GRCh38
rs528992608 Chr.14: 66924243-66924243 on Build GRCh38
rs766752192 Chr.14: 66924328-66924328 on Build GRCh38
rs371948902 Chr.14: 66924184-66924184 on Build GRCh38
rs756588496 Chr.14: 66924205-66924205 on Build GRCh38

View Full Product Details